Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701279

Sigma-Aldrich

Anti-Human IgG (Fc specific), highly cross adsorbed-Biotin antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Goat Anti-Human IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family. It is widely expressed and consists of a gamma heavy chain in the constant (C) region. IgG is a monomer with a molecular weight of about 150 kDa. The structure of IgG constitutes two identical heavy chains and two identical light chains with molecular weight of 50 kDa and 25 kDa respectively. The primary structure of this antibody also contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and resides inside the chains. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4 with different heavy chains, named γ1, γ2, γ3, and γ4, respectively. Limited digestion using papain cleaves the antibody into three fragments, two of which are identical and contain the antigen-binding activity. The third fragment does not possess antigen-binding activity and is known as fragment crystallizable (Fc). It interacts with cells and effector molecules. The Fc fragment contains the CH2 and CH3 domains of the antibody molecule. Maternal IgG is the only antibody transported across the placenta to the fetus. It passively immunizes the infants.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Biotin, Anti-Goat Serum, Human IgG, Human IgG F(c) and Human Serum. No reaction was observed against Human IgG F(ab′)2or Mouse serum proteins.

Immunogen

Human IgG F(c) fragment

Application

Anti-Human IgG (Fc specific)-Biotin antibody produced in goat (highly cross adsorbed) has been used in immunohistochemistry and for binding detection in ligand binding assay.

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibody structure, instability, and formulation
<BIG>Wei W, et al.</BIG>
Journal of Pharmaceutical Sciences (2007)
Human placental Fc receptors and the transmission of antibodies from mother to fetus
<BIG>Neil ES et al.</BIG>
Journal of Reproductive Immunology (1997)
Human Semaphorin 3 Variants Link Melanocortin Circuit Development and Energy Balance
van der Klaauw
Cell null
The structure of a typical antibody molecule
<BIG>Janeway Jr.</BIG>
Antibodies null
Yanfeng Wang et al.
Oncology letters, 14(6), 6857-6862 (2017-11-23)
To investigate Beclin-1 expression in gastric cancer and its clinical relevance, 60 samples were collected from patients with gastric carcinoma, which were subjected to immunohistochemical staining and analysis. Associations of Beclin-1 expression with the clinical parameters of the patients, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service