Skip to Content
Merck
All Photos(2)

Key Documents

SAB3501089

Sigma-Aldrich

Anti-CCR7 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 42 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCR7(1236)

Immunogen

Antibody was raised against a 15 amino acid peptide near the center of human CCR7.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501089.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Peña et al.
AIDS research and human retroviruses, 30(6), 535-541 (2013-11-05)
Mechanisms leading to the observed immune dysregulation in chronic HIV infection are not well understood. The MHC-II class ligand, lymphocyte activation gene-3 (LAG-3, CD223), has been implicated in the complex regulation mechanism of immune functions. In this study, we describe
Reinhold Förster et al.
Nature reviews. Immunology, 8(5), 362-371 (2008-04-02)
A key feature of the immune system is its ability to induce protective immunity against pathogens while maintaining tolerance towards self and innocuous environmental antigens. Recent evidence suggests that by guiding cells to and within lymphoid organs, CC-chemokine receptor 7
M Birkenbach et al.
Journal of virology, 67(4), 2209-2220 (1993-04-01)
Since Epstein-Barr virus (EBV) infection of Burkitt's lymphoma (BL) cells in vitro reproduces many of the activation effects of EBV infection of primary B lymphocytes, mRNAs induced in BL cells have been cloned and identified by subtractive hybridization. Nine genes
R Yoshida et al.
The Journal of biological chemistry, 272(21), 13803-13809 (1997-05-23)
By searching the expressed sequence tag (EST) data base, we identified partial cDNA sequences encoding a novel human CC chemokine. We determined the complete cDNA sequence that encodes a highly basic polypeptide of a total 98 amino acids with 20
Yang Qin et al.
BMC musculoskeletal disorders, 15, 316-316 (2014-09-28)
It is well-documented that both chemokine (C-C motif) ligand 19 (CCL19) and 21 (CCL21) mediate cell migration and angiogenesis in many diseases. However, these ligands' precise pathological role in ankylosing spondylitis (AS) has not been elucidated. The objective of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service