Skip to Content
Merck
All Photos(5)

Key Documents

SAB3500514

Sigma-Aldrich

Anti-ZEB1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AREB6, Anti-BZP, Anti-TCF8, Anti-Zinc finger E-box binding homeobox 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 124 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZEB1(6935)

General description

Zinc finger E-box-binding homeobox 1 (ZEB1) is an EMT (epithelial-mesenchymal transition) transcription factor. The gene encoding this protein is localized on human chromosome 10p11.2.

Immunogen

ZEB1 antibody was raised against a 15 amino acid peptide near the center of human ZEB1.

The immunogen is located within amino acids 530 - 580 of ZEB1.

Application

Anti-ZEB1 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Zinc finger E-box-binding homeobox 1 (ZEB1) controls the expression of E-cadherin as a repressor. It plays an important role in tumor invasion and metastasis. Epigenetic suppression of ZEB1 might be a useful tool for overcoming drug resistance in human breast cancer. Mutations in this gene are associated with posterior polymorphous corneal dystrophy and Fuchs′ endothelial corneal dystrophy (FECD6).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500514.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutational spectrum of the ZEB1 gene in corneal dystrophies supports a genotype-phenotype correlation.
Lechner J
Investigative Ophthalmology & Visual Science (2013)
An hTERT/ZEB1 complex directly regulates E-cadherin to promote epithelial-to-mesenchymal transition (EMT) in colorectal cancer.
Qin Y
Oncotarget (2016)
Novel mutations in the ZEB1 gene identified in Czech and British patients with posterior polymorphous corneal dystrophy.
Liskova P
Human Mutation (2007)
Diminished expression of CRHR2 in human colon cancer promotes tumor growth and EMT via persistent IL-6/Stat3 signaling.
Jorge A
Cellular and molecular gastroenterology and hepatology (2015)
GRHL2-miR-200-ZEB1 maintains the epithelial status of ovarian cancer through transcriptional regulation and histone modification.
Chung VY
Scientific Reports (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service