Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500340

Sigma-Aldrich

Anti-PTK7 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CCK-4, Anti-Colon carcinoma kinase-4, Anti-Protein tyrosine Kinase 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK7(5754)

General description

Protein tyrosine kinase-7 (PTK7) is a transmembrane receptor, that belongs to the receptor protein tyrosine kinase (RTKs) family. It has an extracellular immunoglobulin-like domain, a transmembrane domain and an inert tyrosine kinase-like domain. PTK7 is located on human chromosome 6p21.

Immunogen

PTK7 antibody was raised against a 18 amino acid peptide from near the carboxy terminus of human PTK7.

Application

Anti-PTK7 antibody has been used in immunohistochemistry.

Biochem/physiol Actions

Protein tyrosine kinase-7 (PTK7) stimulates cell proliferation and invasion. It plays a vital role embryonic development and disease. PTK7 controls radioresistance with the help of nuclear factor-κ B in esophageal squamous cell carcinoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500340.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTK7 regulates radioresistance through nuclear factor-kappa B in esophageal squamous cell carcinoma.
Park M, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(10), 14217-14224 (2016)
PTK7 overexpression in colorectal tumors: Clinicopathological correlation and prognosis relevance.
Tian X, et al.
Oncology Reports, 36(4), 1829-1836 (2016)
Ron Chen et al.
Cancer research, 74(10), 2892-2902 (2014-03-22)
Lung cancer remains the most common cause of cancer-related death worldwide and it continues to lack effective treatment. The increasingly large and diverse public databases of lung cancer gene expression constitute a rich source of candidate oncogenic drivers and therapeutic
A meta-analysis of lung cancer gene expression identifies PTK7 as a survival gene in lung adenocarcinoma.
Chen R, Khatri P, Mazur PK, et al.
Cancer Research, 74(10), 2892-2902 (2014)
PTK7 is a novel oncogenic target for esophageal squamous cell carcinoma.
Liu K, et al.
World Journal of Surgical Oncology, 15(1), 105-105 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service