Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500217

Sigma-Aldrich

Anti-Albumin antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Alb, Anti-Serum albumin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALB(213)

Related Categories

Immunogen

Albumin antibody was raised against a 13 amino acid peptide near the center of human Albumin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500217.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxiao Mi et al.
Cellular and molecular gastroenterology and hepatology, 16(5), 657-684 (2023-07-06)
Wilson's disease is an inherited hepatoneurologic disorder caused by mutations in the copper transporter ATP7B. Liver disease from Wilson's disease is one leading cause of cirrhosis in adolescents. Current copper chelators and zinc salt treatments improve hepatic presentations but frequently
A transgenic mouse line for assaying tissue-specific changes in endoplasmic reticulum proteostasis.
Svarcbahs, et al.
Transgenic research, 32, 209-221 (2023)
Shuko Takeda et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(8), 3239-3248 (2013-05-04)
There is a growing body of evidence that soluble oligomeric forms of amyloid β (Aβ) play a critical role in Alzheimer's disease (AD). Despite the importance of soluble Aβ oligomers as a therapeutic target for AD, the dynamic metabolism of
Atanu Paul et al.
Nature communications, 13(1), 930-930 (2022-02-19)
The Hippo/YAP pathway controls cell proliferation through sensing physical and spatial organization of cells. How cell-cell contact is sensed by Hippo signaling is poorly understood. Here, we identified the cell adhesion molecule KIRREL1 as an upstream positive regulator of the
Tianliang Sun et al.
Cell stem cell, 26(1), 97-107 (2019-12-24)
The existence of specialized liver stem cell populations, including AXIN2+ pericentral hepatocytes, that safeguard homeostasis and repair has been controversial. Here, using AXIN2 lineage tracing in BAC-transgenic mice, we confirm the regenerative potential of intestinal stem cells (ISCs) but find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service