Skip to Content
Merck
All Photos(3)

Documents

SAB3500196

Sigma-Aldrich

Anti-CTRP6 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CTRP6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C1q and TNF (Tumour necrosis factor) related protein 6 (CTRP6/C1QTNF6) is a glycoprotein which belongs to family of proteins containing TNF alpha-like globular domain. CTRP6 is a single peptide with a short variable region, a collagen like region, and a C-terminal globular domain. CTRP6 is expressed in human synoviocytes. In human chromosome, the gene is localized on 22q12.3.

Immunogen

CTRP6 antibody was raised against a 15 amino acid peptide from near the carboxy-terminus of human CTRP6.

Biochem/physiol Actions

C1q and TNF (Tumour necrosis factor) related protein 6 (CTRP6) plays a major role in fatty acid metabolism. CTRP6 phosphorylates AMCK (5′-AMP (adenosine monophosphate)-activated protein kinase) and ACC (acetyl coenzyme A carboxylase) and activates AMCK-ACC pathway. The expression level of CTRP6 is increased in rheumatoid arthritis. CTRP6 inhibits alternative complement pathway and helps in maintaining pregnancy. CTRP6 mediates the ectopic lipogenesis in myoblasts through AdipoR1/Erk/PPARγ (Adiponectin receptor 1/Extracellular signal-regulated kinases/Peroxisome proliferator-activated receptor gamma) signalling pathway. CTRP6 is overexpressed in hepatocellular carcinoma and stimulates angiogenesis by Akt (serine/threonine-protein kinase) pathway. CTRP6 confers susceptibility to type I diabetes. CTRP6 maybe a key regulator connecting obesity with tissue inflammation and insulin resistance.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500196.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of a secretory protein C1qTNF6, a C1qTNF family member, in hepatocellular carcinoma
Takeuchi T, et al.
Analytical Cellular Pathology, 34(3), 113-121 (2011)
C1q/TNF-related protein 6 (CTRP6) links obesity to adipose tissue inflammation and insulin resistance
Lei X, et al.
The Journal of Biological Chemistry, 292(36), 14836-14850 (2017)
Lipogenesis in myoblasts and its regulation of CTRP6 by AdipoR1/Erk/PPAR gamma signalling pathway
Wu W, et al.
Acta biochimica et biophysica Sinica, 48(6), 509-519 (2016)
Masanori A Murayama et al.
Nature communications, 6, 8483-8483 (2015-09-26)
The complement system is important for the host defence against infection as well as for the development of inflammatory diseases. Here we show that C1q/TNF-related protein 6 (CTRP6; gene symbol C1qtnf6) expression is elevated in mouse rheumatoid arthritis (RA) models.
Genetic susceptibility to vitiligo: Recent progress from genome-wide association studies
Zhang Z, et al.
Dermatologica Sinica, 32(4), 225-232 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service