Skip to Content
Merck
All Photos(1)

Key Documents

SAB2702227

Sigma-Aldrich

Monoclonal Anti-S Tag antibody produced in mouse

clone GT727, affinity isolated antibody

Synonym(s):

Mouse Anti-KETAAAKFERQHMDS tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT727, monoclonal

form

buffered aqueous solution

concentration

1mg/mL

technique(s)

immunofluorescence: 1:100-1:1,000
western blot: 1:1,000-1:10,000

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

The immunogen used to generate this antibody corresponds to S Tag

Application

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of viral RNAs at ambient temperature via reporter proteins produced through the target-splinted ligation of DNA probes.
Phillips, et al.
Nature Biomedical Engineering (2023)
Mi-Sun Kang et al.
Cell reports, 29(13), 4632-4645 (2019-12-26)
Proliferating cell nuclear antigen (PCNA) is a DNA clamp essential for DNA replication. During DNA synthesis, PCNA is continuously loaded onto and unloaded from DNA. PCNA recruits various proteins to nascent DNA to facilitate chromosome duplication. Therefore, timely PCNA unloading
Sangin Kim et al.
PloS one, 18(5), e0285337-e0285337 (2023-05-19)
Proliferating cell nuclear antigen (PCNA) is a maestro of DNA replication. PCNA forms a homotrimer and interacts with various proteins, such as DNA polymerases, DNA ligase I (LIG1), and flap endonuclease 1 (FEN1) for faithful DNA replication. Here, we identify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service