Skip to Content
Merck
All Photos(1)

Key Documents

SAB2502121

Sigma-Aldrich

Anti-IL15 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

IL-15, MGC9721, OTTHUMP00000164616, OTTHUMP00000164617, interleukin 15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
indirect ELISA: suitable
western blot: suitable

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL15(3600)

General description

Interleukin-15 (IL-15) is an immunomodulating cytokine. The gene encoding it is localized on chromosome 4q31.

Immunogen

Peptide with sequence C-DATLYTESDVHPS, from the internal region of the protein sequence according to NP_000576.1; NP_751915.1

Biochem/physiol Actions

Interleukin-15 (IL-15) stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. It exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. On binding to its receptor, IL-15 is indirectly involved in activating proto-oncogenes. Increased expression of IL-15 has been implicated with rheumatoid arthritis, inflammatory bowel disease, multiple sclerosis and diseases affiliated with retroviruses like human immunodeficiency virus (HIV) and human T-lymphotropic virus-1 (HTLV-I).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromosomal assignment and genomic structure of Il15.
Anderson DM
Genomics, 25(3), 701-706 (1995)
Anjali Mishra et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(8), 2044-2050 (2014-04-17)
Interleukin-15 (IL-15) is a proinflammatory cytokine involved in the development, survival, proliferation, and activation of multiple lymphocyte lineages utilizing a variety of signaling pathways. IL-15 utilizes three distinct receptor chains in at least two different combinations to signal and exert
Huaqin Yuan et al.
Neoplasia (New York, N.Y.), 17(5), 410-420 (2015-05-31)
Intrarenal interleukin-15 (IL-15) plays a major role controlling epithelial survival and polarization both in physiological and pathologic conditions. Herein, we confirmed that human renal cell carcinomas (RCCs) express a membrane-bound IL-15 isoform displaying an unusual molecular weight of 27 kDa.
Patricia K A Mongini et al.
Journal of immunology (Baltimore, Md. : 1950), 195(3), 901-923 (2015-07-03)
Clinical progression of B cell chronic lymphocytic leukemia (B-CLL) reflects the clone's Ag receptor (BCR) and involves stroma-dependent B-CLL growth within lymphoid tissue. Uniformly elevated expression of TLR-9, occasional MYD88 mutations, and BCR specificity for DNA or Ags physically linked
Bieke Broux et al.
Journal of immunology (Baltimore, Md. : 1950), 194(5), 2099-2109 (2015-01-27)
CD4(+)CD28(-) T cells arise through repeated antigenic stimulation and are present in diseased tissues of patients with various autoimmune disorders, including multiple sclerosis (MS). These cells are believed to have cytotoxic properties that contribute to the pathogenic damaging of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service