Skip to Content
Merck
All Photos(2)

Key Documents

SAB2104172

Sigma-Aldrich

Anti-Myelin Basic Protein Antibody

rabbit polyclonal

Synonym(s):

Anti-MGC99675

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
IP
WB
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

Product Name

Anti-MBP antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)

Immunogen

Synthetic peptide directed towards the middle region of human MBP

Sequence

Synthetic peptide located within the following region: FGGDRGAPKRGSGKDSHHPARTAHYGSLPQKSHGRTQDENPVVHFFKNIV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengxi Sun et al.
Plant biotechnology journal, 17(1), 50-62 (2018-05-08)
The optimization of plant architecture in order to breed high-yielding soya bean cultivars is a goal of researchers. Tall plants bearing many long branches are desired, but only modest success in reaching these goals has been achieved. MicroRNA156 (miR156)-SQUAMOSA PROMOTER
Jinxia Hu et al.
FEBS letters, 588(23), 4348-4356 (2014-12-03)
FRMD5 is a novel FERM domain-containing protein depicted in tumor progression. However, the mechanisms underlying FRMD5 inhibition of cell migration is largely unknown. Here, we show that FRMD5 regulates cell migration by interacting with integrin β5 cytoplasmic tail and ROCK1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service