Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411938

Sigma-Aldrich

ANTI-LIMCH1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp434I0312, DKFZp686A01247, DKFZp686B2470, DKFZp686G18243, LIMCH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 101.4 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIMCH1(22998)

Immunogen

LIMCH1 (AAH23546.1, 1 a.a. ~ 897 a.a) full-length human protein.

Sequence
MRKDTDDIESPKRSIRDSGYIDCWDSERSDSLSPPRHGRDDSFDSLDSFGSRSRQTPSPDVVLRGSSDGRGSDSESDLPHRKLPDVKKDDMSARRTSHGEPKSAVPFNQYLPNKSNQTAYVPAPLRKKKAEREEYRKSWSTATSPLGGERPFRYGPRTPVSDDAESTSMFDMRCEEEAAVQPHSRARQEQLQLINNQLREEDDKWQDDLARWKSRRRSVSQDLIKKEEERKKMEKLLAGEDGTSERRKSIKTYREIVQEKERRERELHEAYKNARSQEEAEGILQQYIERFTISEAVLERLEMPKILERSHSTEPNLSSFLNDPNPMKYLRQQSLPPPKFTATVETTIARASVLDTSMSAGSGSPSKTVTPKAVPMLTPKPYSQPKNSQDVLKTFKVDGKVSVNGETVHREEEKERECPTVAPAHSLTKSQMFEGVARVHGSPLELKQDNGSIEINIKKPNSVPQELAATTEKTEPNSQEDKNDGGKSRKGNIELASSEPQHFTTTVTRCSPTVAFVEFPSSPQLKNDVSEEKDQKKPENEMSGKVELVLSQKVVKPKSPEPEATLTFPFLDKMPEANQLHLPNLNSQVDSPSSEKSPVTTPFKFWAWDPEEERRRQEKWQQEQERLLQERYQKEQDKLKEEWEKAQKEVEEEERRYYEEERKIIEDTVVPFTVSSSSADQLSTSSSMTEGSGTMNKIDLGNCQDEKQDRRWKKSFQGDDSDLLLKTRESDRLEEKGSLTEGALAHSGNPVSKGVHEDHQLDTEAGAPHCGTNPQLAQDPSQNQQTSNPTHSSEDVKPKTLPLDKSINHQIESPSERRKSISGKKLCSSCGLPLGKGAAMIIETLNLYFHIQCFRCGICKGQLGDAVSGTDVRIRNGLLNCNDCYMRSRSAGQPTTL

Biochem/physiol Actions

LIMCH1 (LIM and calponin homology domains 1) is a multidomain protein belonging to the human mosaic domain superfamily. It is composed of calponin homology (CH) domain that acts as actin binding site and the conserved cysteine rich double Zn finger possessing Lim domain. Both the domain exist as a singlet. It has been reported that these proteins may exist in several duplex and quadruplet CH containing forms. It may be involved in the cytoskeletal and signaling pathways.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix Friedberg
Molecular biology reports, 36(5), 1059-1081 (2008-06-17)
In this paper we examine (restricted to homo sapiens) the products resulting from gene duplication and the subsequent alternative splicing for the members of a multidomain group of proteins which possess the evolutionary conserved calponin homology CH domain, i.e. an
T Stradal et al.
FEBS letters, 431(2), 134-137 (1998-08-26)
A sequence motif of about 100 amino acids, termed the 'calponin homology domain' has been suggested to confer actin binding to a variety of cytoskeletal and signalling molecules. Here we analyse and compare the sequences of all calponin homology domain-containing
Felix Friedberg
Molecular biology reports, 37(4), 1707-1716 (2009-07-01)
In this paper, the inventory presented for singlet CH (calponin homology/actin binding) domain containing human multidomain proteins is extended to several duplex and one quadruplet CH containing forms. Invariably, the duplexes are located at the begin of the molecules. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service