Skip to Content
Merck
All Photos(1)

Documents

SAB1411364

Sigma-Aldrich

Anti-ACTG2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ACT, ACTA3, ACTE, ACTL3, ACTSG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41.9 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTG2(72)

General description

Actins are highly conserved proteins that are involved in various types of cell motility, and maintenance of the cytoskeleton. In vertebrates, three main groups of actin isoforms, alpha, beta and gamma have been identified. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. Actin, gamma 2, encoded by this gene, is a smooth muscle actin found in enteric tissues. (provided by RefSeq)

Immunogen

ACTG2 (NP_001606.1, 1 a.a. ~ 376 a.a) full-length human protein.

Sequence
MCEEETTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKPEYDEAGPSIVHRKCF

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui-Rui Chen et al.
Chemico-biological interactions, 302, 11-21 (2019-02-01)
Angiotensin II-related cardiac fibrosis is one of the key pathological changes of the hypertrophied left ventricle in various heart disease. Irisin was recently reported to confer cardio-protective and anti-oxidative effects, while whether it can reverse the renin-angiotensin-aldosterone system(RAAS) activation related(angiotensin
Qinghui Xiong et al.
Pharmacological research, 144, 90-98 (2019-04-09)
Hydrogen sulfide gas (H2S) has protective effects in the cardiovascular system that includes preventing the development of atherosclerosis when tested in several in vivo models. Plaque instability is a major risk factor for thromboembolism, myocardial infarction, and stroke, so we
Heinz P Janke et al.
Advanced healthcare materials, 7(18), e1800605-e1800605 (2018-07-31)
In vivo monitoring of tissue-engineered constructs is important to assess their integrity, remodeling, and degradation. However, this is challenging when the contrast with neighboring tissues is low, necessitating labeling with contrast agents (CAs), but current CAs have limitations (i.e., toxicity
Isabelle Matte et al.
BMC cancer, 19(1), 406-406 (2019-05-02)
CA125 is a well-established ovarian cancer (OC) serum biomarker. The CA125 heavily glycosylated epitope is carried by the MUC16 mucin, a high molecular weight transmembrane mucin. Upon proteolytic cleavage, the extracellular domain of MUC16 is released from the cell surface
Yingnan Bai et al.
Journal of thoracic disease, 11(5), 1849-1859 (2019-07-10)
The regulation of angiogenesis in the treatment of cardiovascular diseases has been widely studied and the vascular endothelial growth factor (VEGF) families and VEGF receptor (VEGFR) have been proven to be one of the key regulators. The VEGFR endocytosis has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service