Skip to Content
Merck
All Photos(2)

Documents

SAB1410844

Sigma-Aldrich

Anti-NNMT antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 29.6 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NNMT(4837)

Related Categories

General description

The nicotinamide N-methyltransferase (NNMT) gene, spanning 16.5kb of genomic DNA with three exons and two introns, is mapped to human chromosome 11q23.1. The encoded protein consists of 264 amino acids with a predicted molecular mass of 29.6kDa. NNMT is expressed in cytoplasm.

Immunogen

NNMT (NP_006160.1, 1 a.a. ~ 264 a.a) full-length human protein.

Sequence
MESGFTSKDTYLSHFNPRDYLEKYYKFGSRHSAESQILKHLLKNLFKIFCLDGVKGDLLIDIGSGPTIYQLLSACESFKEIVVTDYSDQNLQELEKWLKKEPEAFDWSPVVTYVCDLEGNRVKGPEKEEKLRQAVKQVLKCDVTQSQPLGAVPLPPADCVLSTLCLDAACPDLPTYCRALRNLGSLLKPGGFLVIMDALKSSYYMIGEQKFSSLPLGREAVEAAVKEAGYTIEWFEVISQSYSSTMANNEGLFSLVARKLSRPL

Biochem/physiol Actions

Nicotinamide N-methyltransferase (NNMT) is a cytosolic methyltransferase enzyme. It catalyzes the N-methylation of nicotinamide, pyridines and other structural analogs. Hence, it plays a vital role in the biotransformation and detoxification of various xenobiotic compounds. Altered expression of the gene is associated with the pathogenesis of various diseases such as acute lymphoblastic leukemia (ALL), parkinson′s disease, thyroid cancer, gastric cancer, colorectal cancer, lung, liver and oral carcinomas. In addition, variation in the gene might increase the risk of susceptibility to hyperlipidemia and migraine.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicotinamide-N-Methyltransferase gene rs694539 variant and migraine risk.
Sazci A
The Journal of Headache and Pain, 17 (2016)
Physiological Study on Association between Nicotinamide N-Methyltransferase Gene Polymorphisms and Hyperlipidemia.
Zhu XJ
BioMed Research International (2016)
NNMT (nicotinamide N-methyltransferase)
Emanuelli M
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service