Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410813

Sigma-Aldrich

Anti-MYOD1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MYF3, MYOD, PUM, bHLHc1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 34.5 kDa

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYOD1(4654)

General description

Myogenic differentiation 1 (MYOD1) is encoded by the gene mapped to human chromosome 11p15.1. The encoded protein belongs to the basic helix-loop-helix (bHLH) family of transcription factors. MYOD1 is expressed exclusively during normal skeletal muscle myogenesis and in rhabdomyosarcomas.

Immunogen

MYOD1 (NP_002469.2, 1 a.a. ~ 320 a.a) full-length human protein.

Sequence
MELLSPPLRDVDLTAPDGSLCSFATTDDFYDDPCFDSPDLRFFEDLDPRLMHVGALLKPEEHSHFPAAVHPAPGAREDEHVRAPSGHHQAGRCLLWACKACKRKTTNADRRKAATMRERRRLSKVNEAFETLKRCTSSNPNQRLPKVEILRNAIRYIEGLQALLRDQDAAPPGAAAAFYAPGPLPPGRGGEHYSGDSDASSPRSNCSDGMMDYSGPPSGARRRNCYEGAYYNEAPSEPRPGKSAAVSSLDCLSSIVERISTESPAAPALLLADVPSESPPRRQEAAAPSEGESSGDPTQSPDAAPQCPAGANPNPIYQVL

Biochem/physiol Actions

Myogenic differentiation 1 (MYOD1) associates with histone-modifying enzymes and stimulates muscle differentiation by elevating the expression of histone H3 mono methyl K4 (H3K4me1) and histone H3 acetyl 27 (H3K27ac). MyoD1 staining can be considered as a key adjunct in the sorting of pediatric soft-tissue sarcomas. Mutation in the gene is associated with the pathogenesis of perinatally lethal fetal akinesia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A recurrent neomorphic mutation in MYOD1 defines a clinically aggressive subset of embryonal rhabdomyosarcoma associated with PI3K-AKT pathway mutations.
Kohsaka S
Nature Genetics, 46, 595-600 (2014)
Deficiency of the myogenic factor MyoD causes a perinatally lethal fetal akinesia.
Watson CM
Journal of medical Genetics, 53, 264-269 (2016)
Myogenic regulatory protein (MyoD1) expression in childhood solid tumors: diagnostic utility in rhabdomyosarcoma.
Dias P
The American Journal of Pathology, 137, 1283-1291 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service