Skip to Content
Merck
All Photos(3)

Key Documents

SAB1405796

Sigma-Aldrich

Anti-FASN antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FAS, MGC14367, MGC15706, OA-519, SDR27X1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~48.3 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FASN(2194)

General description

FASN (fatty acid synthase) encodes a fatty acid synthase (FAS) enzyme. This gene is located on human chromosome 17.
The enzyme encoded by this gene is a multifunctional protein. Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA, in the presence of NADPH, into long-chain saturated fatty acids. In some cancer cell lines, this protein has been found to be fused with estrogen receptor-α (ER-α), in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-α. (provided by RefSeq)

Immunogen

FASN (AAH07909.1, 1 a.a. ~ 439 a.a) full-length human protein.

Sequence
MSTNDTIVSGTLPQRMASCLEVLDLFLNQPHMVLSSFVLAEKAAAYRDRDSQRDLVEAVAHILGIRDLAAVNLDSSLADLGLDSLMSVEVRQTLERELNLVLSVREVRQLTLRKLQELSSKADEASELACPTPKEDGLAQQQTQLNLRSLLVNPEGPTLMRLNSVQSSERPLFLVHPIEGSTTVFHSLASRLSIPTYGLQCTRAAPLDSIHSLAAYYIDCIRQVQPEGPYRVAGYSYGACVAFEMCSQLQAQQSPAPTHNSLFLFDGSPTYVLAYTQSYRAKLTPGCEAEAETEAICFFVQQFTDMEHNRVLEALLPLKGLEERVAAAVDLIIKSHQGLDRQELSFAARSFYYKLRAAEQYTPKAKYHGNVMLLRAKTGGAYGEDLGADYNLSQVCDGKVSVHVIEGDHRTLLEGSGLESIISIIHSSLAEPRVSVREG

Biochem/physiol Actions

FASN (fatty acid synthase) is essential for de novo fatty-acid synthesis. It plays a major role in glioma neovascularization. The product of this gene helps in the production of fatty acids from acetyl-CoA and malonyl-CoA. Absence of FASN may block migration, invasion and healing of hepatoma carcinoma cells by reducing HIF-1α (hypoxia-inducible factor 1-α) and IGFBP1 (insulin-like growth factor binding protein-1).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of FASN suppresses migration, invasion and growth in hepatoma carcinoma cells by deregulating the HIF-1?/IGFBP1 pathway
Gong J, et al.
International Journal of Oncology, 50(3), 883-892 (2017)
Effects of fatty acid synthase inhibitors on lymphatic vessels: an in vitro and in vivo study in a melanoma model.
Bastos DC, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 97(2), 194-194 (2017)
FASN, dietary fat intake, and risk of uterine leiomyomata in the Black Women's Health Study
Wise LA, et al.
Fertility and Sterility, 106(5), 1136-1141 (2016)
Inhibition of fatty acid synthase suppresses neovascularization via regulating the expression of VEGF-A in glioma
Zhou Y, et al.
Journal of Cancer Research and Clinical Oncology, 142(12), 2447-2459 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service