Skip to Content
Merck
All Photos(4)

Key Documents

SAB1405590

Sigma-Aldrich

Anti-CD44 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDW44, CSPG8, ECMR-III, HCELL, IN, LHR, MC56, MDU2, MDU3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~39.4 kDa

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

General description

The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). This protein participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis. (provided by RefSeq)

Immunogen

CD44 (ENSP00000263398, 1 a.a. ~ 361 a.a) full-length human protein.

Sequence
MDKFWWHAAWGLCLVPLSLAQIDLNITCRFAGVFHVEKNGRYSISRTEAADLCKAFNSTLPTMAQMEKALSIGFETCRYGFIEGHVVIPRIHPNSICAANNTGVYILTSNTSQYDTYCFNASAPPEEDCTSVTDLPNAFDGPITITIVNRDGTRYVQKGEYRTNPEDIYPSNPTDDDVSSGSSSERSSTSGGYIFYTFSTVHPIPDEDSPWITDSTDRIPATRDQDTFHPSGGSHTTHGSESDGHSHGSQEGGANTTSGPIRTPQIPEWLIILASLLALALILAVCIAVNSRRRCGQKKKLVINSGNGAVEDRKPSGLNGEASKSQEMVHLVNKESSETPDQFMTADETRNLQNVDMKIGV

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Jubber et al.
Disease models & mechanisms, 16(7) (2023-06-09)
Little is known about the distal excretory component of the urinary tract in Danio rerio (zebrafish). This component is affected by many human diseases and disorders of development. Here, we have undertaken multi-level analyses to determine the structure and composition
Guo-Fang Guan et al.
Oncology letters, 10(2), 631-636 (2015-12-02)
According to the cancer stem cell theory, a small subpopulation of cancer cells, known as cancer stem cells (CSCs), exist that are self-renewing and are involved in tumor invasion, metastasis and recurrence. A number of studies have reported that certain
Ayman I Elkady
American journal of translational research, 11(4), 2550-2569 (2019-05-21)
Prostate cancer is still at the forefront causes of cancer-related morbidity and mortality in men throughout the globe. The disease is initiated and fostered by a subset of cancer stem cells (CSCs). Costus speciosus is an oriental herb used in
Fahad Paryani et al.
Nature communications, 15(1), 6742-6742 (2024-08-08)
The mechanisms underlying the selective regional vulnerability to neurodegeneration in Huntington's disease (HD) have not been fully defined. To explore the role of astrocytes in this phenomenon, we used single-nucleus and bulk RNAseq, lipidomics, HTT gene CAG repeat-length measurements, and
Juan Zhou et al.
OncoTargets and therapy, 12, 7067-7077 (2019-10-01)
Src homology 2-containing inositol 5-phosphatase 2 (SHIP2) gene is associated with arthrosclerosis, gastric cancer and diabetes. In this study, we revealed that overexpression of SHIP2 is closely implicated with the development of breast cancer (BC). The BC tissue and adjacent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service