Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402110

Sigma-Aldrich

Monoclonal Anti-NR0B1, (C-terminal) antibody produced in mouse

clone 1F10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

AHC, AHCH, AHX, DAX-1, DAX1, DSS, GTD, HHG, NROB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
direct immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR0B1(190)

General description

This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism. (provided by RefSeq)

Immunogen

NR0B1 (NP_000466, 361 a.a. ~ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
IKCFLSKCWSLNISTKEYAYLKGTVLFNPDVPGLQCVKYIQGLQWGTQQILSEHTRMTHQGPHDRFIELNSTLFLLRFINANVIAELFFRPIIGTVSMDDMMLEMLCTKI

Biochem/physiol Actions

The encoded orphan nuclear receptor is involved in the growth and activity of the adrenal and reproductive organs. It is significantly associated with the growth of gonads, development of sexual characteristics and steroidogenesis. The expression of NR0B1 (nuclear receptor subfamily 0 group B member 1) gene is regulated by Nanog (transcription factor). NR0B1 is known to control pluripotency and differentiation of embryonic stem cells in mice. Mutation in NR0B1 is importantly associated with X-linked adrenal hypoplasia congenita. Mutations in NR0B1 is also known to downregulate the androgen receptor activity, also leading to defect in spermatogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-linked adrenal hypoplasia congenita and hypogonadotropic hypogonadism: Identification and in vitro study of a novel small indel in the NR0B1 gene.
Yu T
Molecular Medicine Reports, 13(5), 4039-4045 (2016)
DAX1 Overexpression in Pediatric Adrenocortical Tumors: A Synergic Role with SF1 in Tumorigenesis.
de Sousa GR
Hormone and Metabolic Research, 47(9), 656-661 (2015)
A Novel Mutation of DAX-1 Associated with Secretory Azoospermia.
Mou L
PLoS ONE, 10(7), 1-11 (2015)
A novel DAX-1 mutation in two male siblings presenting with precocious puberty and late-onset hypogonadotropic hypogonadism.
Liu Y
Journal of Pediatric Endocrinology & Metabolism : JPEM, 30(3), 349-353 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service