Skip to Content
Merck
All Photos(6)

Key Documents

SAB1301290

Sigma-Aldrich

ANTI-LC3 (APG8C) (N-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

LC3C, MAP1LC3C, microtubule-associated protein 1 light chain 3 gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

16852 Da

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyou-Chan Choi et al.
Journal of neurochemistry, 112(2), 366-376 (2009-11-03)
Our previous microarray analysis identified a neuroprotective protein Oxi-alpha, that was down-regulated during oxidative stress (OS)-induced cell death in dopamine neurons [Neurochem. Res. (2004) vol. 29, pp. 1223]. Here we find that the phylogenetically conserved Oxi-alpha protects against OS by
Anita Thyagarajan et al.
Nutrition and cancer, 62(5), 630-640 (2010-06-25)
Medicinal mushroom Ganoderma lucidum is one of the most esteemed natural products that have been used in the traditional Chinese medicine. In this article, we demonstrate that G. lucidum triterpene extract (GLT) suppresses proliferation of human colon cancer cells HT-29
Guanghong Liao et al.
Brain research, 1270, 140-151 (2009-03-31)
Niemann-Pick Type C (NPC) disease is a devastating developmental disorder with progressive and fatal neurodegeneration. Previous work has shown that a single injection of the neurosteroid allopregnanolone at postnatal day 7 significantly prolonged lifespan of Npc1-/- mice. However, the cellular/molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service