Skip to Content
Merck
All Photos(3)

Documents

S9571

Sigma-Aldrich

Anti-SUMO-2 antibody produced in rabbit

enhanced validation

~0.6 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SMT3B, Anti-SMT3H2, Anti-Sentrin-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~15 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~0.6 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using HeLa cells
western blot: 0.5-1 μg/mL using nuclear extract of HeLa cells or cell extract of HEK293-T cells transfected with human SUMO-2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SUMO2(6613)
mouse ... Sumo2(170930)

General description

Small ubiquitin-related modifier (SUMO-2) gene is mapped to human chromosome 17q25.1. Sumo-2 is related to SUMO-3 with a 86% sequence identity. It comprises a sumoylation motif at the residue lysine 11.

Specificity

Anti-SUMO-2 recognizes human SUMO-2.

Immunogen

synthetic peptide corresponding to amino acids 80-93 located at the C-terminal of human SUMO-2, conjugated to KLH. This sequence is identical in many species including rat, mouse, pig, and bovine SUMO-2 and is identical in human SUMO-3.

Application

Anti-SUMO-2 antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Small ubiquitin-related modifier (SUMO-2) is essential for the cellular defense in hyperthermic cytotoxicity. Along with SUMO-3, SUMO2 is essential for cell survival post-heat shock. SUMO-2 has the ability to exist as self-modified polymers. SUMO-2 and SUMO-3 negatively regulate interferon induction.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 1% bovien serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Lin Su et al.
Gene, 296(1-2), 65-73 (2002-10-18)
The SUMO (small ubiquitin-like modifier) protein and ubiquitin have similar 3-D structure. Sumolyzation and ubiquitination exhibit similar biological processes for post-translational modification. However, unlike ubiquitination, which targets proteins for degradation, sumolyzation participates in a number of cellular processes such as
John T Crowl et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(26), 6798-6803 (2018-06-13)
Detection of nucleic acids by innate immune sensors triggers the production of type I interferons (IFNs). While IFNs are essential for host defense against viral infection, dysregulated production of IFNs underlies numerous autoinflammatory diseases. We have found that the loss
Rajarshi Chakrabarti et al.
Biochemical and biophysical research communications, 439(2), 209-214 (2013-09-03)
Phosphatidylinositol 4 phosphate 5 kinase 1α (PIP5K) is mainly localized in the cytosol and plasma membrane. Studies have also indicated its prominent association with nuclear speckles. The exact nature of this nuclear pool of PIP5K is not clear. Using biochemical
Filip Golebiowski et al.
Science signaling, 2(72), ra24-ra24 (2009-05-28)
Covalent conjugation of the small ubiquitin-like modifier (SUMO) proteins to target proteins regulates many important eukaryotic cellular mechanisms. Although the molecular consequences of the conjugation of SUMO proteins are relatively well understood, little is known about the cellular signals that
A B Steffensen et al.
Acta physiologica (Oxford, England), 222(3) (2017-09-10)
The voltage-gated potassium channel KV 11.1 is the molecular basis for the IKr current, which plays an important role in cardiac physiology. Its malfunction is associated with both inherited and acquired cardiac arrhythmias. Native currents differ from those in experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service