S8776
Sodium chloride solution
0.9% in water, BioXtra, suitable for cell culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75
grade:
for analytical purposes
Recommended Products
grade
for analytical purposes
Quality Level
sterility
sterile-filtered
product line
BioXtra
concentration
0.9% in water
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
SMILES string
[Na+].[Cl-]
InChI
1S/ClH.Na/h1H;/q;+1/p-1
InChI key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Sodium chloride is widely used in biochemistry and molecular biology research. It is a component of phosphate buffered saline (Product No. P3813) and βC buffer (Product Nos. S0902 and S8015). Sodium chloride is a component of all media formulations. This product has been qualified for use in preparation of in vitro solutions.
Application
Sodium chloride solution has been used:
- for intraperitoneal administration in mice to investigate the effect of toll-like receptors (TLR-3) activation on neonatal hypoxic-ischemic (HI)
- to dissolve bleomycin sulfate for bleomycin treatments in mice
- to dilute tribromoethanol for intraperitoneal injection in mice
Packaging
PET bottle (100 ml), PETG bottle (20 ml), with caps
Preparation Note
Prepared in tissue culture grade water.
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Koji Tanaka et al.
Nature communications, 6, 6337-6337 (2015-02-27)
Pore-forming toxins (PFT) are water-soluble proteins that possess the remarkable ability to self-assemble on the membrane of target cells, where they form pores causing cell damage. Here, we elucidate the mechanism of action of the haemolytic protein fragaceatoxin C (FraC)
Laurent Dortet et al.
Journal of clinical microbiology, 52(7), 2359-2364 (2014-04-25)
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in
Frédéric Lebrun-Julien et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(25), 8432-8448 (2014-06-21)
The mammalian target of rapamycin (mTOR) pathway integrates multiple signals and regulates crucial cell functions via the molecular complexes mTORC1 and mTORC2. These complexes are functionally dependent on their raptor (mTORC1) or rictor (mTORC2) subunits. mTOR has been associated with
Luc H Boudreau et al.
Blood, 124(14), 2173-2183 (2014-08-02)
Mitochondrial DNA (mtDNA) is a highly potent inflammatory trigger and is reportedly found outside the cells in blood in various pathologies. Platelets are abundant in blood where they promote hemostasis. Although lacking a nucleus, platelets contain functional mitochondria. On activation
Articles
Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service