Skip to Content
Merck

S8281

Safflower seed oil from Carthamus tinctorius seed

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
EC Number:
232-276-5
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant seeds (Carthamus tinctorius)

Quality Segment

form

liquid

refractive index

n20/D 1.476 (lit.)

density

0.921 g/mL at 25 °C (lit.)

functional group

carboxylic acid, oleic acid

lipid type

oils

shipped in

ambient

storage temp.

room temp

Application

<ul>
<li><strong>Anti-osteoporosis effects of mammalian lignans and their precursors from flaxseed and safflower seed using zebrafish model:</strong> The study investigates the potential health benefits of safflower seed oil in preventing osteoporosis, highlighting its utility as a dietary supplement (Yang et al., 2023).</li>
</ul>

Physical form

Raw oil without preservative


Storage Class

10 - Combustible liquids

wgk

awg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library