Skip to Content
Merck
All Photos(1)

Key Documents

S6510

Sigma-Aldrich

N-Succinyl-Leu-Leu-Val-Tyr-7-Amido-4-Methylcoumarin

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H53N5O10
CAS Number:
Molecular Weight:
763.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

0.1% trifluoroacetic acid in acetonitrile: water (3:1): 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)CCC(O)=O)C(=O)NC(CC(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc1ccc(O)cc1)C(=O)Nc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C40H53N5O10/c1-21(2)16-29(42-33(47)14-15-34(48)49)38(52)43-30(17-22(3)4)39(53)45-36(23(5)6)40(54)44-31(19-25-8-11-27(46)12-9-25)37(51)41-26-10-13-28-24(7)18-35(50)55-32(28)20-26/h8-13,18,20-23,29-31,36,46H,14-17,19H2,1-7H3,(H,41,51)(H,42,47)(H,43,52)(H,44,54)(H,45,53)(H,48,49)

InChI key

UVFAEQZFLBGVRM-UHFFFAOYSA-N

Amino Acid Sequence

N-Suc-Leu-Leu-Val-Tyr-7-AMC

Application

N-succinyl-Leu-Leu-Val-Tyr-7-amido-4-methylcoumarin was used in proteasome chymotrypsin-like activity assay in Jurkat cell lysate6 and crude cell lysate of rice.7

Biochem/physiol Actions

In the presence of chymotrypsin-like enzyme activity, the fluorophore, 7-amido-4-methylcoumarin is released from N-succinyl-Leu-Leu-Val-Tyr-7-amido-4-methylcoumarin. The fluorescence obtained is a measure of the enzyme activity.6

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Fluorogenic substrate for chymotrypsin-like enzymes, such as cathepsin B and calpain which have been implicated in programmed cell death.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Izaltina Silva-Jardim et al.
Acta tropica, 91(2), 121-130 (2004-07-06)
Proteasomes are multisubunit proteases that exist universally among eukaryotes. They have multiple proteolytic activities and are believed to have important roles in regulating cell cycle, selective intracellular proteolysis, and antigen presentation. Here we have partially purified Leishmania chagasi proteasome. The
Turnover of oxidatively damaged nuclear proteins in BV-2 microglial cells is linked to their activation state by poly-ADP-ribose polymerase.
O Ullrich et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(8), 1460-1462 (2001-06-02)
T Reinheckel et al.
The Biochemical journal, 335 ( Pt 3), 637-642 (1998-10-31)
Oxidatively modified ferritin is selectively recognized and degraded by the 20S proteasome. Concentrations of hydrogen peroxide (H2O2) higher than 10 micromol/mg of protein are able to prevent proteolytic degradation. Exposure of the protease to high amounts of oxidants (H2O2, peroxynitrite
R Guerra-Sá et al.
Experimental parasitology, 109(4), 228-236 (2005-03-10)
Proteasomes are multi-subunit proteases involved in several mechanisms and thought to contribute to the regulation of cellular homeostasis. Here, we report for the first time biochemical evidence for the existence of a ubiquitin-proteasome proteolytic pathway in this parasite. Proteasomes from
S Glockzin et al.
The Journal of biological chemistry, 274(28), 19581-19586 (1999-07-03)
The ubiquitin/proteasome pathway mediates the degradation of many short-lived proteins that are critically involved in the regulation of cell proliferation and cell death, including the tumor suppressor protein p53. Accumulation of p53 and induction of apoptosis in RAW 264.7 macrophages

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service