Skip to Content
Merck

Skip To

S5007

Sunflower seed oil

from Helianthus annuus, liquid, emollient

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
EC Number:
232-273-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sunflower seed oil from Helianthus annuus,

density

0.92 g/mL at 25 °C (lit.)

Quality Segment

General description

Sunflower seed oil from Helianthus annuus contains saturated fatty acids (palmitic and stearic acids), monounsaturated fatty acid (oleic acid), and polyunsaturated acid (mostly linoleic acid).[1] In addition, this edible oil also contains nonglyceridic contents including tocopherols and plant sterols, and is rich in vitamins and minerals.

Application

Sunflower seed oil from Helianthus annuus was used as a vehicle to administer tamoxifen[2][3] and 4-hydroxytamoxifen[4] to mice.

Biochem/physiol Actions

Sunflower seed oil is one of the most used vegetable oils in human nutrition.[1] Oilseed crop sunflower (Helianthus annuus) is also used for biodiesel production. Sunflower oil is used as an emollient in cosmetics, and it serves as an effective phenolic antioxidant. Phytosterols in sunflower oil help in regulating the cholesterol levels in the body. Therefore, sunflower oil may be used to treat pathological conditions such as like acne, arthritis, and hair damage.[5]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2989471231642347
Quality Level

100

Quality Level

300

Quality Level

100

Quality Level

-

density

0.92 g/mL at 25 °C (lit.)

density

-

density

-

density

-


Still not finding the right product?


supp_hazards

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters