Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.32
UNSPSC Code:
41106500
Pricing and availability is not currently available.
storage temp.
room temp
Quality Segment
Application
Spin Columns are recommended for use with Seppro™ reagents.
Legal Information
Seppro is a trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level - | Quality Level 100 |
| storage temp. room temp | storage temp. 2-8°C | storage temp. 20-25°C | storage temp. - |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



