Skip to Content
Merck
All Photos(1)

Key Documents

S4562

Sigma-Aldrich

β-Secretase inhibitor

≥97% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H118N16O27
Molecular Weight:
1651.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥97% (HPLC)

form

powder

mol wt

~_1.65 kDa

color

white

mp

200 °C

solubility

DMSO: soluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

N([C@H]([C@@H](O)CC(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H](Cc1ccccc1)C(=O)O)CC(C)C)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)[C@H](O)C)CCC(=O)O)C

InChI

1S/C73H118N16O27/c1-11-37(8)59(88-66(108)45(23-27-56(101)102)79-63(105)43(21-25-54(97)98)81-72(114)60(39(10)91)89-62(104)41(75)19-15-16-28-74)71(113)85-49(33-90)68(110)80-44(22-26-55(99)100)65(107)87-58(36(6)7)70(112)83-47(31-51(76)93)67(109)82-46(29-34(2

InChI key

CQTBDEGWLSDJEP-JMAXQNTPSA-N

Amino Acid Sequence

Lys-Thr-Glu-Glu-Ile-Ser-Glu-Val-Asn-Sta-Val-Ala-Glu-Phe

General description

β-Site amyloid precursor protein (APP) cleaving enzyme-1 (BACE1), an aspartic protease belongs to the protease family of enzymes comprises of six luminal cysteine residues. These residues help in the formation of three intermolecular disulfide bonds and N-linked glycosylation sites.

Application

β-Secretase inhibitor has been used as β-site amyloid precursor protein (APP) cleaving enzyme-1 (BACE1) inhibitor in BACE1 inhibitor assay.(2)
β-Secretase inhibitor may be used to inhibit BACE1 in studies related to AD.

Biochem/physiol Actions

β-Site amyloid precursor protein (APP) cleaving enzyme-1 (BACE1) is a β-secretase that initiates the production of amyloid protein in Alzheimer′s dieases (AD) patients. β-Secretase inhibitors decrease the generation of β amyloid and formation of amyloid plaques typical of AD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

beta-secretase inhibitor; a promising novel therapeutic drug in Alzheimer?s disease
Menting KW, et al.
Frontiers in Aging Neuroscience, 6, 165-165 (2014)
Hans Hilpert et al.
Journal of medicinal chemistry, 56(10), 3980-3995 (2013-04-18)
An extensive fluorine scan of 1,3-oxazines revealed the power of fluorine(s) to lower the pKa and thereby dramatically change the pharmacological profile of this class of BACE1 inhibitors. The CF3 substituted oxazine 89, a potent and highly brain penetrant BACE1
Jungmi Lee et al.
Analytica chimica acta, 1022, 89-95 (2018-05-08)
Amyloid-β (Aβ) is generated by proteolytic processing of amyloid precursor protein (APP) by beta-secretase (BACE-1) and gamma-secretase. Amyloid-β is responsible for the formation of senile plaques in Alzheimer's disease (AD). Consequently, inhibition of β-secretase (BACE-1), a rate-limiting enzyme in the
B J Baranowski et al.
Physiological reports, 12(16), e70001-e70001 (2024-08-20)
Brain-derived neurotrophic factor (BDNF) content and signaling has been identified as one potential regulator of amyloid precursor protein (APP) processing. Recently published work has demonstrated that BDNF reduces BACE1 activity while also elevating the inhibition of GSK3β in the prefrontal
Yukai Wang et al.
Biomolecules, 13(12) (2023-12-23)
Nrg1 (Neuregulin 1) type III, a susceptible gene of schizophrenia, exhibits a critical role in the central nervous system and is essential at each stage of Schwann's cell development. Nrg1 type III comprises double-pass transmembrane domains, with the N-terminal and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service