Skip to Content
Merck
All Photos(1)

Key Documents

S3644

Sigma-Aldrich

Spectrin from human erythrocytes

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human erythrocytes

Quality Level

form

buffered aqueous glycerol solution

UniProt accession no.

application(s)

cell analysis

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SPTA1(6708)

General description

Spectrin is the major component of the protein network, which covers the cytoplasmic surface of vertebrate erythrocyte membranes. It is a high molecular weight heterodimer composed of two subunits (molecular weights of approximately 230 kDa and 250 kDa). Spectrins are extended flexible molecules approximately 200-260 nm in length and 3-6 nm across with actin-binding domains at each end. Spectrins are composed of α and β subunits, which are both related to a-actinin. The α and β subunits associate laterally to form antiparallel heterodimers and the heterodimers are assembled head-head to form heterotetramers. The erythrocyte membrane skeleton is organized as a polygonal network formed by five to seven extended spectrin molecules linked to short actin filaments approximately 40 nm in length. The spectrin-actin network of erythrocytes is coupled to the membrane bilayer primarily through the association of spectrin with ankyrin, which in turn is bound to the cytoplasmic domain of the anion exchanger. A major function of the spectrin skeleton in erythrocytes is to provide mechanical support for the membrane bilayer and allow survival of the cells in circulation.
The C-terminus of spectrin contains a calmodulin-like EF domain. This domain is critical for binding of spectrin to actin for proper cytoskeletal formation. Deletion of this domain results in weak, irregularly-shaped red blood cells.

Quality

Highly purified

Physical form

Solution in 50% glycerol containing 100 mM sodium chloride, 10 mM phosphate buffer, pH 8.0, 1 mM dithiothreitol, 1mM EDTA and 0.1 mM phenylmethylsulfonyl fluoride.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew N Rasband
Current biology : CB, 23(5), R197-R198 (2013-03-12)
Axons must be supported by a strong and flexible cytoskeleton. New 'super-resolution' imaging of the submembranous axonal cytoskeleton reveals that it is organized in a periodic, ladder-like structure with alternating rings of actin linked together by intervening complexes of spectrin.
Matthew D Vesely et al.
Annals of the New York Academy of Sciences, 1284, 1-5 (2013-05-09)
Accumulated data from animal models and human cancer patients strongly support the concept that the immune system can identify and control nascent tumor cells in a process called cancer immunosurveillance. In addition, the immune system can also promote tumor progression
Pan Zhang et al.
Nucleic acids research, 41(10), 5321-5340 (2013-04-11)
Telomere integrity is critical for telomere function and genomic stability. We previously demonstrated that non-erythroid α-spectrin (αIISp) is present in mammalian cell nuclei where it is important in repair of DNA interstrand cross-links (ICLs) and chromosome stability. We now demonstrate
Peter Bellstedt et al.
Journal of biomolecular NMR, 54(4), 325-335 (2012-11-28)
We have carried out chemical shift correlation experiments with symmetry-based mixing sequences at high MAS frequencies and examined different strategies to simultaneously acquire 3D correlation spectra that are commonly required in the structural studies of proteins. The potential of numerically
Laura Picas et al.
ACS nano, 7(2), 1054-1063 (2013-01-26)
The erythrocyte membrane, a metabolically regulated active structure that comprises lipid molecules, junctional complexes, and the spectrin network, enables the cell to undergo large passive deformations when passing through the microvascular system. Here we use atomic force microscopy (AFM) imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service