Skip to Content
Merck

S3452

s-TDM Adjuvant

oil-in-water emulsion

Synonym(s):

6’-Dicorynomycolate, sTDM, TDCM, Trehalose-6, sTDCM, synthetic cord factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12161804
Clone:
-
Species reactivity:
-
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

shipped in

ambient

storage temp.

2-8°C

Biochem/physiol Actions

s-TDM Adjuvant is a stable oil-in-water emulsion which can be used as an alternative to the classical Freund′s water-in-oil emulsion or as a boost for the Sigma Adjuvant System S6322.

Physical form

Each vial contains 0.5 mg synthetic trehalose dicorynomycolate (an analogue of trehalose dimycolate from the cord factor of the tubercle bacillus) in 44 μL of squalene oil, 0.2% TWEEN® 80 and water.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library