Skip to Content
Merck
All Photos(1)

Documents

S193

Sigma-Aldrich

Anti-Synapsin I antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-EPILX, Anti-MRX50, Anti-SYN1a, Anti-SYN1b, Anti-SYNI

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen ~80 kDa (Synapsin Ia)

species reactivity

rat, mouse, human, bovine

technique(s)

dot blot: 1:1000
immunohistochemistry (frozen sections): 1:2000
immunoprecipitation (IP): 1 μg
indirect immunofluorescence: 1:2000
western blot: 1:1000

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYN1(6853)
mouse ... Syn1(20964)
rat ... Syn1(24949)

General description

Synapsin I (1a and 1b) refers to two nearly identical phosphoproteins found in the cytoplasmic surface of synaptic vesicles of the CNS (central nervous system) and PNS(peripheral nervous system). It may be used to localize and detect synapsin I (synapsins Ia and Ib are collectively referred to as synapsin I) in nerve terminals.The SYN1 gene is mapped to human chromosome Xp11.3-p11.23.

Immunogen

synapsin I from bovine brain.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.


Rabbit polyclonal anti-Synapsin I antibody can be used for the localization and detection of synapsin I (synapsins Ia and Ib, approximately 80 kDa and 77 kDa, are collectively referred to as synapsin I) in nerve terminals.

Biochem/physiol Actions

Synapsin I is involved in the regulation of axonogenesis and synaptogenesis wherein it serves as a substrate of various protein kinases including those activated by cAMP, calciuim/calmodulin, mitogens, and cyclins. The antibody strongly reacts with the synapsin I doublet. Synapsin I is associated with vesicle trafficking. Mutation in this gene is one of the known cause for familial epilepsy and autism spectrum disorders.

Physical form

Lyophilized from ammonium bicarbonate (5 mM)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synapsin III: role in neuronal plasticity and disease.
Porton B, et al.
Seminars in Cell & Developmental Biology, 22(4), 416-424 (2011)
Nonsense-mediated mRNA decay and loss-of-function of the protein underlie the X-linked epilepsy associated with the W356? mutation in synapsin I.
Giannandrea M, et al.
PLoS ONE, 8(6), e67724-e67724 (2013)
DNA hypomethylation of Synapsin II CpG islands associates with increased gene expression in bipolar disorder and major depression.
Cruceanu C, et al.
BMC Psychiatry, 16(1), 286-286 (2016)
PRICKLE1 interaction with SYNAPSIN I reveals a role in autism spectrum disorders.
Paemka L, et al.
PLoS ONE, 8(12), e80737-e80737 (2013)
Andrew San Antonio et al.
The Journal of comparative neurology, 522(6), 1333-1354 (2013-10-30)
The hippocampal CA2 subfield was initially identified by Lorente de Nó as an anatomically distinct region based on its cytoarchitectural features. Although there is an enormous body of literature on other hippocampal subfields (CA1 and CA3), relatively little is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service