Skip to Content
Merck
All Photos(1)

Documents

RES1458C

SAFC

CDAP

Synonym(s):

1-Cyano-4-dimethylaminopyridinium tetrafluoroborate, CDAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BF4N3
CAS Number:
Molecular Weight:
234.99
Beilstein:
5685616
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Quality Level

storage temp.

−20°C

SMILES string

[F-].FB(F)F.CN(C)c1cc[n+](cc1)C#N

InChI

1S/C8H10N3.BF3.FH/c1-10(2)8-3-5-11(7-9)6-4-8;2-1(3)4;/h3-6H,1-2H3;;1H/q+1;;/p-1

InChI key

ONCRRSYBEJHQMM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Packaging

Product is available in the following package sizes:
RES1458C-B101X: 100 mg container
RES1458C-B102X: 500 mg container
RES1458C-B103X: 1 g container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Hwa Choi et al.
Clinical and vaccine immunology : CVI, 23(2), 162-167 (2015-12-18)
The efficacy of the serotype 3 (ST3) pneumococcal conjugate vaccine (PCV) remains unclear. While the synthesis of capsular polysaccharide (CPS) of most serotypes is wzy dependent, the strains of two serotypes, 3 and 37, synthesize CPS by the synthase-dependent pathway
HyunChul Youn et al.
Journal of affective disorders, 239, 30-36 (2018-07-12)
Depression is a risk factor for mild cognitive impairment (MCI) and for the conversion from MCI to Alzheimer's disease (AD). This study investigated regional cerebral glucose metabolism (rCMglc) in older adults with depression and MCI, either with or without amyloidopathy.
Song Chen et al.
Acta pharmacologica Sinica, 30(7), 1008-1014 (2009-07-04)
YCP, a novel (1,4)-alpha-D-glucan, was isolated from the mycelium of the marine filamentous fungus Phoma herbarum YS4108. In this work, we investigated a YCP-binding cellular receptor expressed by macrophages and the intracellular signal transduction pathways involved in YCP-induced macrophage activation.
Ting Li et al.
International journal of biological macromolecules, 133, 1194-1200 (2019-04-15)
The effects of a polysaccharide (DAP) from the roots of Dipsacus asper on renal complication in streptozotocin (STZ)-induced diabetic rats were investigated in the present study. Administration of DAP (100 and 300 mg/kg) for 4 weeks significantly decreased the fasting blood glucose
Valerian B Pinto et al.
PloS one, 8(11), e79304-e79304 (2013-11-19)
A native outer membrane vesicles (NOMV) vaccine was developed from three antigenically diverse strains of Neisseria meningitidis that express the L1,8, L2, and L3,7 lipooligosaccharide (LOS) immunotypes, and whose synX, and lpxL1 genes were deleted.. Immunogenicity studies in mice showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service