Skip to Content
Merck

RAB1365

Human CHL-1 ELISA

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... CHL1(10752)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CHL1(10752)

General description

This ELISA antibody pair detects Human Cell Adhesion Molecule with Homology to L1CAM (L1CAM-2)
CHL1 (cell adhesion molecule L1 like) gene codes for a cell-adhesion molecule. It belongs to the L1 family of cell adhesion molecules. The CHL1gene is mapped to human chromosome 3p26.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

The axon guidance protein encoded by CHL1 (cell adhesion molecule L1 like) gene plays a major role in the guidance of thalamocortical axons and the multiplication and differentiation of neural progenitor cells. CHL1 acts as a tumor-suppressor gene in breast cancer (BC). Mutation in the axon guidance protein results in horizontal gaze palsy with progressive scoliosis (HGPPS). It participates in mental development.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library