RAB1049
Human Fibronectin ELISA Kit
for cell culture supernatants, plasma, and serum samples
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.6 ng/ml
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... FN1(2335)
General description
This ELISA antibody pair detects human Fibronectin. Other species are not determined.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amanda Munasinghe et al.
Cancer letters, 477, 88-96 (2020-03-03)
Pancreatic ductal adenocarcinoma (PDAC) is one of the deadliest of all solid tumours and more effective therapy is urgently needed. The stroma is thought to play a critical role in tumour development and metastasis, and high stromal expression of the
Zhonghua Lv et al.
OncoTargets and therapy, 13, 6443-6452 (2020-09-05)
S100A9, which is expressed in prostate cancer, has been reported in association with prostate cancer progression. However, the role of S100A9 in prostate cancer metastasis is largely unknown. The aim of this study was to investigate the effect of S100A9
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service