Skip to Content
Merck
All Photos(2)

Key Documents

RAB0183

Sigma-Aldrich

Human bFGF ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 102.4-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF2(2247)

General description

The Human bFGF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human bFGF cell lysate and tissue lysate.

Immunogen

Recombinant Human basic-FGF

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohua Liu et al.
Psychiatry research, 218(1-2), 54-60 (2014-05-06)
Multiple neurotrophic and/or growth factors, recently nominated as "angioneurins", play the key roles in mood modulation and neuroplasticity, and their dysfunction might be involved in the pathophysiology and treatment of mood disorders. We examined serum levels of vascular endothelial growth
In Kyong Shim et al.
Journal of biomedical materials research. Part A, 102(10), 3639-3648 (2013-11-23)
Efficient osseointegration is a key factor in dental implants to reduce the total time-course of therapy. Titanium implants with anodized surface gained much interest for their enhanced osseointegration. Anodized implant combined with bioactive drugs is an ideal candidate for enhance
Kyung Min Park et al.
Biomacromolecules, 15(6), 1979-1984 (2014-04-18)
Many women around the world are suffering from urinary incontinence, defined as the unintentional leakage of urine by external abnormal pressure. Although various kinds of materials have been utilized to treat this disease, therapies that are more effective are still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service