Skip to Content
Merck
All Photos(1)

Key Documents

R9281

Sigma-Aldrich

Rotigotine hydrochloride

≥98% (HPLC)

Synonym(s):

(−)-(S)-5,6,7,8-Tetrahydro-6-(propyl(2-(2-thienyl)ethyl)amino)-1-naphthol hydrochloride, (−)-N 0437, N 0923, Neupro, SPM 962

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NOS·HCl
CAS Number:
Molecular Weight:
351.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

UCB Inc.

storage temp.

room temp

SMILES string

Cl.CCCN(CCc1cccs1)[C@H]2CCc3c(O)cccc3C2

InChI

1S/C19H25NOS.ClH/c1-2-11-20(12-10-17-6-4-13-22-17)16-8-9-18-15(14-16)5-3-7-19(18)21;/h3-7,13,16,21H,2,8-12,14H2,1H3;1H/t16-;/m0./s1

InChI key

CEXBONHIOKGWNU-NTISSMGPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Rotigotine hydrochloride is a non-ergolinic D3/D2 and D1-dopamine agonist. It is appropriate for transdermal delivery through skin patches, that has the drug in a silicone-based adhesive matrix.
Rotigotine is a dopamine receptor agonist with preference for D3 receptors over D2 and D1. As such, it is an effective anti-Parkinsonian agent. Racemic rotigotine is about 50 times as potent as quinpirole, the gold standard D2 agonist.

Features and Benefits

This compound was developed by UCB Inc.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikos Gorgoraptis et al.
Brain : a journal of neurology, 135(Pt 8), 2478-2491 (2012-07-05)
Hemispatial neglect following right-hemisphere stroke is a common and disabling disorder, for which there is currently no effective pharmacological treatment. Dopamine agonists have been shown to play a role in selective attention and working memory, two core cognitive components of
Santiago Perez-Lloret et al.
Fundamental & clinical pharmacology, 27(1), 81-95 (2012-02-11)
Rotigotine, a non-ergot dopamine agonist, has been developed as a novel transdermal formulation. The rotigotine transdermal patch has received EMEA marketing authorization for the treatment of adult patients with early or advanced Parkinson's disease (PD) or with moderate to severe
Lawrence W Elmer et al.
Parkinsonism & related disorders, 18(5), 488-493 (2012-02-14)
This prospective, open-label extension (SP702; NCT00594165) of a 6-month double-blind, randomized study investigated the long-term safety and tolerability of rotigotine transdermal system in early Parkinson's disease (PD). Patients with early-stage idiopathic PD received transdermal rotigotine for up to 6 years
Jan-Peer Elshoff et al.
Clinical therapeutics, 34(4), 966-978 (2012-03-10)
The dopamine agonist rotigotine is formulated in a transdermal delivery system (patch) for once-daily application. It has been reported as efficacious in the treatment of idiopathic Parkinson's disease (PD) and restless legs syndrome. This article summarizes the results of 3
Heike Beneš et al.
Sleep medicine, 13(6), 589-597 (2012-04-17)
To assess the risk of augmentation under treatment with the transdermally delivered dopamine agonist rotigotine for restless legs syndrome (RLS). Experts in RLS augmentation retrospectively reviewed data from two double-blind, placebo-controlled 6-month trials (745 rotigotine and 214 placebo subjects, NCT00136045

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service