Skip to Content
Merck

R6000

Ribonuclease S from bovine pancreas

Grade XII-S

Synonym(s):

RNase S, pancreatic RNase, protease-modified RNase A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-646-6
MDL number:
Specific activity:
≥60 Kunitz units/mg solid
Biological source:
bovine pancreas
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas

Quality Segment

type

Grade XII-S

form

powder

specific activity

≥60 Kunitz units/mg solid

mol wt

13.7 kDa

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

Application

Ribonuclease S from bovine pancreas has been used in a study to assess the hybridase activity of human ribonuclease-1. Ribonuclease S from bovine pancreas has also been used in a study to investigate a new approach to obtaining high-activity RNase, DNase, cholesterolesterase, and trypsin from cattle pancreas.

Other Notes

Protease-modified RNase A


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library