Skip to Content
Merck
All Photos(3)

Key Documents

R101

Sigma-Aldrich

Ranitidine hydrochloride

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N4O3S · HCl
CAS Number:
Molecular Weight:
350.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

tan

solubility

H2O: 1.8 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 7.0 mg/mL

originator

GlaxoSmithKline

SMILES string

Cl[H].CN\C(NCCSCc1ccc(CN(C)C)o1)=C\[N+]([O-])=O

InChI

1S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15H,6-8,10H2,1-3H3;1H/b13-9-;

InChI key

GGWBHVILAJZWKJ-CHHCPSLASA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

Application

Ranitidine hydrochloride has been used as a reference compound for the development and validation of parallel artificial membrane permeability assay (PAMPA).

Biochem/physiol Actions

H2 histamine receptor antagonist; anti-ulcer agent.
Ranitidine is mainly used to treat gastrointestinal impairment instigated by non-steroidal anti-inflammatory drugs (NSAIDs).

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P W Bliss et al.
Alimentary pharmacology & therapeutics, 13(12), 1669-1674 (1999-12-14)
Gastrin release by Helicobacter pylori may be an important step in the pathway leading to duodenal ulceration. A histamine H3-receptor agonist was found to release gastrin from antral mucosal fragments; this was interpreted as being due to suppression of somatostatin
Ying Liu et al.
Molecular and cellular biochemistry, 448(1-2), 61-69 (2018-02-07)
Abnormal angiogenesis is critically involved in tumor progression and metastasis including endometrial cancer and is regulated by microRNAs such as microRNA-101 (miR-101). We hypothesize that miR-101 expression is disrupted in endometrial cancer and modulation of miR-101 levels is sufficient to
Tian Xie et al.
Neuroscience research, 158, 30-36 (2019-09-19)
We aimed to demonstrate the effects of microRNA (miR)-101 on neuropathic pain and explore the underlying mechanisms. Rat spinal microglia cells were isolated and inflammatory condition was stimulated by 24-h incubation with lipopolysaccharide (LPS). Rats were divided into 4 groups:
Ranitidine
Grant S M, et al.
Drugs, 37(6), 801-870 (1989)
Ruth Prieto-Montero et al.
Sensors (Basel, Switzerland), 20(19) (2020-10-03)
Functionalized fluorescent silica nanoparticles were designed and synthesized to selectively target cancer cells for bioimaging analysis. The synthesis method and characterization of functionalized fluorescent silica nanoparticles (50-60 nm), as well as internalization and subcellular localization in HeLa cells is reported

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service