Skip to Content
Merck
All Photos(3)

Key Documents

R0878

Sigma-Aldrich

D-Ribulose 1,5-bisphosphate sodium salt hydrate

≥90% (TLC)

Synonym(s):

D-erythro-2-Pentulose, 1,5-bis(dihydrogen phosphate), D-Ribulose 1,5-diphosphate, RuDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12O11P2 · xNa+ · yH2O
CAS Number:
Molecular Weight:
310.09 (anhydrous free acid basis)
UNSPSC Code:
12352201
NACRES:
NA.28

biological source

synthetic (inorganic)

Quality Level

Assay

≥90% (TLC)

form

powder

impurities

≤16% water (Karl Fischer)

color

white

solubility

water: ~50 g/L

cation traces

Na: 17.1-24.6% (dry basis)

storage temp.

−20°C

SMILES string

[P](=O)(OC[C@@H](O)[C@@H](O)C(=O)CO[P](=O)(O)O)(O)O

InChI

1S/C5H12O11P2/c6-3(1-15-17(9,10)11)5(8)4(7)2-16-18(12,13)14/h3,5-6,8H,1-2H2,(H2,9,10,11)(H2,12,13,14)/t3-,5-/m1/s1

InChI key

YAHZABJORDUQGO-NQXXGFSBSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Bio-Inspired Microreactors Continuously Synthesize Glucose Precursor from CO(2) with an Energy Conversion Efficiency 3.3 Times of Rice.: This study leverages D-Ribulose 1,5-bisphosphate sodium salt hydrate in bio-inspired microreactors to synthesize glucose precursors from CO₂. The approach demonstrates an energy conversion efficiency significantly higher than that of rice, highlighting its potential for efficient carbon fixation and bioengineering applications (Zhu et al., 2024).

  • Designing Stacked Assembly of Type III Rubisco for CO₂ Fixation with Higher Efficiency.: The research utilizes D-Ribulose 1,5-bisphosphate sodium salt hydrate to enhance the efficiency of CO₂ fixation through the stacked assembly of Type III Rubisco. This innovative approach offers improvements in metabolic engineering for carbon capture and utilization (Zeng et al., 2022).

  • Continuous artificial synthesis of glucose precursor using enzyme-immobilized microfluidic reactors.: This study explores the continuous synthesis of glucose precursors using microfluidic reactors with immobilized enzymes, including D-Ribulose 1,5-bisphosphate sodium salt hydrate. The technique demonstrates potential for scalable biochemical production processes (Zhu et al., 2019).

Biochem/physiol Actions

Ribulose-1,5-bisphosphate (RuBP) is a component of the Calvin cycle that is metabolized into glycerate 3-phosphate (G3P) by the enzyme ribulose bisphosphate carboxylase/oxygenase (RuBisCO). RuBP is used to identify, differentiate and characterized ribulose bisphosphate carboxylase(s)/oxygenase(s) (RuBisCO).

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert E Sharwood et al.
Journal of experimental botany, 67(10), 3137-3148 (2016-04-29)
Plants operating C3 and C4 photosynthetic pathways exhibit differences in leaf anatomy and photosynthetic carbon fixation biochemistry. Fully understanding this underpinning biochemical variation is requisite to identifying solutions for improving photosynthetic efficiency and growth. Here we refine assay methods for
S Ogushi et al.
The Journal of biological chemistry, 265(19), 10943-10949 (1990-07-05)
The tissue contents of previously known allosteric effectors of brain phosphofructokinase (EC 2.7.1.11) (PFK) and the kinetic behavior of isolated PFK were investigated during the initiation of rapid glycolytic flux in freeze-blown rat brain. Comparing 0- with 5-s brains revealed
M Sawada et al.
The international journal of biochemistry & cell biology, 32(4), 447-454 (2000-04-13)
6-Phosphofructo-1-kinase and fructose-1,6-bisphosphatase are rate-limiting enzymes for glycolysis and gluconeogenesis respectively, in the fructose 6-phosphate/fructose 1,6-bisphosphate cycle in the liver. The effect of ribose 1,5-bisphosphate on the enzymes was investigated. Ribose 1,5-bisphosphate synergistically relieved the ATP inhibition and increased the
I Ozaki et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 125(1), 97-102 (2000-06-07)
Fructose-1,6-bisphosphatase is one of the regulatory enzymes of gluconeogenesis in kidney cortex. The effect of ribose 1,5-bisphosphate on fructose-1,6-bisphosphatase purified from rat kidney cortex was studied. Rat kidney cortex, fructose-1,6-bisphosphatase exhibited hyperbolic kinetics with regard to its substrate, but the
T Kawaguchi et al.
The Journal of biological chemistry, 276(30), 28554-28561 (2001-05-25)
Macrophages can adapt to the absence of oxygen by switching to anaerobic glycolysis. In this study, we investigated (a) the roles of fructose 2,6-bisphosphate (Fru-2,6-P2) and ribose 1,5-bisphosphate (Rib-1,5-P2), potent activators of phosphofructokinase, (b) the enzymes responsible for the synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service