Skip to Content
Merck
All Photos(1)

Key Documents

PZ0123

Sigma-Aldrich

Avridine

≥97% (HPLC)

Synonym(s):

CP 20961, N,N-Dioctadecyl-N′,N′-bis(2-hydroxyethyl)-1,3-diaminopropane, N,N-Dioctadecyl-N′,N′-bis(2-hydroxyethyl)propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H90N2O2
CAS Number:
Molecular Weight:
667.19
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥5 mg/mL (with warming to 60 °C for 5 minutes)

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCCN(CCCCCCCCCCCCCCCCCC)CCCN(CCO)CCO

InChI

1S/C43H90N2O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-36-44(38-35-39-45(40-42-46)41-43-47)37-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h46-47H,3-43H2,1-2H3

InChI key

WXNRAKRZUCLRBP-UHFFFAOYSA-N

Biochem/physiol Actions

Avridine is a potent synthetic adjuvant that can induce arthritis in most rat strains; immunomodulator and interferon-inducing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic factors regulating experimental arthritis in mice and rats.
R L Wilder et al.
Current directions in autoimmunity, 1, 121-165 (2002-01-17)
A E Kingston et al.
Clinical and experimental immunology, 103(1), 77-82 (1996-01-01)
The effects of a mycobacterial 71-kD hsp antigen have been investigated for its ability to modulate arthritis in rats. Subcutaneous injection (base of tail) of increasing amounts of hsp71 from Mycobacterium tuberculosis (MTB) produced dose-dependent differential inhibitory effects on induction
A Wigdorovitz et al.
Vaccine, 16(17), 1627-1632 (1998-08-26)
We have previously demonstrated that the presence of the antigen presenting cells (APC) is critical in the induction and maintenance of the immune response in animals infected or immunized with inactivated FMDV. The use of immunological adjuvants has been repeatedly
A M Sadir et al.
Veterinary immunology and immunopathology, 69(1), 11-22 (1999-09-18)
The epidemiological analysis of the cattle population during the eradication plan of foot and mouth disease (FMD) in Argentina clearly indicated a higher incidence of the disease in animals within their first year of age. It is important to improve
D M Pérez Filgueira et al.
Vaccine, 17(4), 345-352 (1999-02-13)
Different immunomodulators have been previously tested in our laboratory as enhancers of the specific immune response to FMDV vaccines in a murine model [2-4]. Here, we present results of two of these immunomodulators, a water-soluble fraction of the cell wall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service