Skip to Content
Merck
All Photos(2)

Key Documents

PRS3197

Sigma-Aldrich

Anti-Caspase-12 (small) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Caspase 12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 46 kDa

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Casp12(12364)

General description

Caspase-12 (CASP12) is an immunomodulatory factor, that is present in the cytoplasmic side of the ER (endoplasmic reticulum). It is upregulated highly in muscle, liver and kidney.

Immunogen

a synthetic peptide corresponding to amino acids within the small cleavage product of murine caspase-12.

Biochem/physiol Actions

Caspase-12 (CASP12) plays an important role in endoplasmic reticulum (ER)-related apoptosis. It moderates hepatocyte apoptosis prompted by carbon tetrachloride. Lack of caspase-12 (CASP12) increases cytokine responses.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3197.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caspase-12 deficiency enhances cytokine responses but does not protect against lethal Plasmodium yoelii 17XL infection
Miu J, et al.
Parasite Immunology, 32(11-12), 773-778 (2010)
Caspase-12 mediates carbon tetrachloride-induced hepatocyte apoptosis in mice
Liu H, et al.
World Journal of Gastroenterology, 20(48), 18189-18198 (2014)
The resveratrol attenuates ethanol-induced hepatocyte apoptosis via inhibiting ER-related caspase-12 activation and PDE activity in vitro
Liu LQ, et al.
Alcoholism, Clinical and Experimental Research, 38(3), 683-693 (2014)
Caspase-12, but Not Caspase-11, Inhibits Obesity and Insulin Resistance
Skeldon AM, et al.
Journal of Immunology, 196(1), 437-447 (2016)
Juan Carlos Gómora-García et al.
Cell death discovery, 7(1), 131-131 (2021-06-05)
Altered protein homeostasis is associated with neurodegenerative diseases and acute brain injury induced under energy depletion conditions such as ischemia. The accumulation of damaged or unfolded proteins triggers the unfolded protein response (UPR), which can act as a homeostatic response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service