Skip to Content
Merck

PLA0230

Rabbit anti-Beta-catenin Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

88kDa, CTNNB, MRD19, armadillo, beta 1, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, IHC, IP, WB
Citations:
6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

ChIP: 4-30 μg, immunohistochemistry: 1:100- 1:500, immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_001895.1

application(s)

research pathology

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Beta-catenin(1499)

Immunogen

The epitope recognized by PLA0230 maps to a region between residue 760 and 781 of human beta-catenin using the numbering given in entry NP_001895.1 (GeneID 1499).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Beta-catenin plays an integral role in the Wnt signaling pathway and is critical to morphogenic signaling and body axis specification. Beta-catenin exists in complexes with cadherins, transmembrane proteins that mediate cell adhesion. When Wnt-signaling proteins bind their receptors, beta-catenin is stabilized and accumulates in the cell. Translocation of stabilized beta-catenin allows its association with transcription factors and the activation of specific gene expression.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library