Skip to Content
Merck

PHYTOMLS01

Phytochemical Metabolite Library Of Standards Plate 1 (Ethanol Soluble)

Supplied by IROA Technologies

Synonym(s):

Mass Spectrometry Metabolite Library, PHYTOMLS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

packaging

pkg of 1 plates

manufacturer/tradename

IROA Technologies

drug control

Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

PHYTOMLS Plate 1 is a collection of high quality small biochemical molecules that span a broad range of primary metabolism. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays and phenotypic screening. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID. The library is intended to be used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.

Application

Metabolite qualification and quantification for MS, NMR, etc.; Functional cellular assays; Phenotypic screening
Suitable for manual and automated work flows

Features and Benefits

To view a complete list of compounds in the library visit PHYTOMLS Plate Map. The complete plate map with name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMBD ID, PubChem Compound and Substance ID are provided with the software dowmload. Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds: Carboxylic acids, amino acids; Biogenic amines, polyamines; Nucleotides, coenzymes and vitamins; Mono-and disaccharides; Fatty acids, lipids, steroids and hormones

Preparation Note

PHYTOMLS Plate 1 compounds are conveniently provided at 5μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on PHYTOMLS01, solubilization and preparation notes please refer to the product information sheet at the top of the page.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
MLSDiscovery software package is distributed with and is tailored to work with PHYTOMLS01. A user manual and video instructions are provided.

To download the software package, visit sigma.com/mlsdiscovery.

To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.

Legal Information

PHYTOMLS is a trademark of IROA Technologies LLC


signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 2 Inhalation - STOT SE 3

target_organs

Lungs, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library