Skip to Content
Merck
All Photos(1)

Key Documents

P9256

Sigma-Aldrich

Psychosine from bovine brain

lyophilized powder, ≥98% (TLC)

Synonym(s):

1-β-D-Galactosylsphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H47NO7
CAS Number:
Molecular Weight:
461.63
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

lyophilized powder

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C24H47NO7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(27)18(25)17-31-24-23(30)22(29)21(28)20(16-26)32-24/h14-15,18-24,26-30H,2-13,16-17,25H2,1H3/b15-14+/t18-,19+,20+,21-,22-,23+,24+/m0/s1

InChI key

HHJTWTPUPVQKNA-PIIMIWFASA-N

Application

Psychosine has been used as a lipid in lipid-protein overlay binding assay.

Biochem/physiol Actions

Psychosine is a neurotoxic lysosphingolipid and a glycolipid precursor for the synthesis of cerebrosides. It is synthesized from galactosylceramide, phrenosin. Elevated psychosine levels are implicated in Krabbe disease. It mediates demyelination and consequently oligodendrocytes degeneration. Psychosine is also correlated to neurological disease, Globoid cell leukodystrophy (GLD), where in galactosylceramidase (GALC) enzyme deficiency leads to its accumulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Benjamin Smith et al.
ASN neuro, 3(4), doi:10-doi:10 (2011-09-21)
Infantile Krabbe disease results in the accumulation of lipid-raft-associated galactosylsphingosine (psychosine), demyelination, neurodegeneration and premature death. Recently, axonopathy has been depicted as a contributing factor in the progression of neurodegeneration in the Twitcher mouse, a bona fide mouse model of
Wenjin Li et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 1503-1512 (2020-07-14)
Metachromatic leukodystrophy (MLD) is a rare genetic disease characterised by a dysfunction of the enzyme arylsulphatase A leading to the lysosomal accumulation of cerebroside sulphate (sulphatide) causing subsequent demyelination in patients. The enzyme galactosylceramide (cerebroside) sulphotransferase (CST) catalyses the transfer
Conserved roles of C. elegans and human MANFs in sulfatide binding and cytoprotection
Bai M, et al.
Nature Communications, 9(1), 897-897 (2018)
Hiroshi Yamamoto et al.
Archives of biochemistry and biophysics, 506(1), 83-91 (2010-11-18)
Sphingolipids act as signaling mediators that regulate a diverse range of cellular events. Although numerous sphingolipid functions have been studied, little is known about the effect of sphingolipids on monocyte differentiation into macrophages. Here, we report that two lysosphingolipids, sphingosylphosphorylcholine
Lysosphingolipids and sphingolipidoses: Psychosine in Krabbe's disease
Spassieva S and Bieberich E
Journal of Neuroscience Research, 94(11), 974-981 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service