Skip to Content
Merck
All Photos(1)

Documents

P8996

Sigma-Aldrich

Monoclonal Anti-PINCH-1 antibody produced in mouse

clone PINCH-C58, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PINCH-C58, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

hamster, bovine, mouse, monkey, canine, human, rat

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using total cell extract from 293T cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LIMS1(3987)
mouse ... Lims1(110829)
rat ... Gcc2(309798)

General description

Monoclonal Anti-PINCH-1 (mouse IgG1 isotype) is derived from the hybridoma PINCH-C58 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a peptide of human PINCH1, conjugated to KLH. LIM and senescent cell antigen-like-containing domain protein 1 (LIMS1) or particularly interesting new Cys-His protein 1 (PINCH-1) is widely expressed. It has five LIM domains. It has leucine-rich nuclear export signal (NES) and nuclear localization signal (NLS) at the C-terminus. Human PINCH-1 and 2 are encoded by two different genes that are 82% identical in their amino acid sequence.

Immunogen

peptide corresponding to amino acids 281-299 of human PINCH-1, conjugated to KLH.

Application

Monoclonal Anti-PINCH-1 antibody produced in mouse has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunoprecipitation
  • co-immunoprecipitation

Biochem/physiol Actions

The interaction with PINCH proteins (PINCH-1 and 2) is important for localization of integrin linked kinase protein to cell matrix contact sites. It plays a key role in cell−matrix adhesion. PINCH2 is co-expressed and co-localized with PINCH-1/ILK complex. PINCH1 interacts with Wilms tumor 1 protein (WT1) in podocytes. Elevated expression of PINCH1 is observed in human laryngeal carcinoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PINCH: More than just an adaptor protein in cellular response
Kovalevich J, et al.
Journal of Cellular Physiology, 226(4), 940-947 (2011)
Characterization of PINCH-2, a new focal adhesion protein that regulates the PINCH-1-ILK interaction, cell spreading, and migration
Zhang Y, et al.
The Journal of biological chemistry, 277(41), 38328-38338 (2002)
High PINCH1 Expression in Human Laryngeal Carcinoma Associates with Poor Prognosis
Tsinias G, et al.
Analytical Cellular Pathology, 2018 (2018)
PINCH1 is transcriptional regulator in podocytes that interacts with WT1 and represses podocalyxin expression
Wang D, et al.
Testing, 6(2), e17048-e17048 (2011)
Iveta Dobreva et al.
Journal of proteome research, 7(4), 1740-1749 (2008-03-11)
Protein-protein interactions play an essential role in the regulation of vital biological functions. Through a network of interactions, integrin-linked kinase (ILK) functions downstream of integrin receptors to control cell spreading, migration, growth, survival, and cell cycle progression. Despite many reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service