Skip to Content
Merck

P8491

Nunclon® MicroWell plates for automation

96 well (with lid), round bottom clear wells

Synonym(s):

automation 96 well plate, Nunc 163320, cell cultivation plate, culture plate, multiwell plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
clear polystyrene , polystyrene , round bottom clear wells
Size:
96 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear polystyrene , polystyrene , round bottom clear wells

sterility

sterile

feature

lid

packaging

pack of 50 (individually wrapped)

manufacturer/tradename

Nunc 163320

maximum volume

300 μL

size

96 wells

well maximum volume

300 μm

working volume

250 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

General description

  • Standard 96 well footprint meets the SBS guidelines
  • High Flange ideal for robotics and automation
  • Nunclon Delta hydrophilic surface that promotes cell attachment and growthP
  • Features a tight closing lid that ensures sample protection and prevents evaporation
  • Raised rim minimizes cross-contamination risk
  • Stackable - A convenient space-saving option
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
Nunc® MicroWell 96-well microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Plates are high quality for consistent and reliable results.

Application

  • Antibiotic screens
  • Cell-based assays
  • Screening compounds.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library