Skip to Content
Merck
All Photos(1)

Key Documents

P8170

Sigma-Aldrich

Protein G−Peroxidase from Streptococcus sp.

recombinant, expressed in unspecified host, lyophilized powder

Synonym(s):

Protein G−HRP from Streptococcus sp.

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

recombinant

expressed in unspecified host

Quality Level

conjugate

peroxidase conjugate

form

lyophilized powder

technique(s)

direct ELISA: 1:100,000 using human IgG

storage temp.

−20°C

General description

Protein G is a bacterial cell wall protein, which has high affinity for immunoglobulin G (IgG). It is isolated from group G streptococcal strains.[1] Peroxidase from Streptococcus sp. is used to maintain the H2O2 homeostasis in cells. It aids the regulation of ABC Mn2+-permease complex (psaBCA) genes.[2]

Specificity

The product binds IgG for most mammalian species (excluding feline) and can be used in ELISA or immunohistochemical formats. Protein was determined to be at 250-350ug per 250ug vial by UV absorbance. Reactive to chicken IgG as well.

Application

Protein G- Peroxidase from Streptococcus sp. has been used in indirect ELISA and immunoblotting.[3][4]

Preparation Note

Labeled with peroxidase type VI by a modification of the procedure of O′Sullivan, M.S., et al., FEBS Lett., 95, 311 (1978) which favors low molecular weight conjugates. Conjugate is purified by gel filtration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Thiol peroxidase is an important component of Streptococcus pneumoniae in oxygenated environments
Hajaj B, et al.
Infection and Immunity, IAI-00126 (2012)
Evaluation of a modified Rose Bengal test and an indirect enzyme-linked immunosorbent assay for the diagnosis of Brucella melitensis infection in sheep
Ferreira AC, et al.
Veterninary Research, 34(3), 297-305 (2003)
Dimitra Zarafeta et al.
Frontiers in microbiology, 7, 1779-1779 (2016-12-03)
Lipolytic enzymes that retain high levels of catalytic activity when exposed to a variety of denaturing conditions are of high importance for a number of biotechnological applications. In this study, we aimed to identify new lipolytic enzymes, which are highly
P Nol et al.
Clinical and vaccine immunology : CVI, 16(3), 323-329 (2009-01-09)
Monitoring of the kinetics of production of serum antibodies to multiple mycobacterial antigens can be useful as a diagnostic tool for the detection of Mycobacterium bovis infection as well as for the characterization of disease progression and the efficacy of
Santos B S A S et al.
RSC advances, 12(49), 31711-31728 (2022-11-17)
The current COVID-19 pandemic presents one of the greatest challenges in human history. There is a consensus that the rapid and accurate diagnosis of COVID-19 directly affects procedures to avoid dissemination, promote treatments, and favor the prognosis of infected patients.

Protocols

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

See All

Questions

  1. After diluting this, how long does it keep in a fridge? (2-8 degrees Celsius)

    1 answer
    1. The solution should be stable for about one week when stored at 2-8°C. For long-term storage, the stock solution should be frozen in aliquots at -20°C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service