Skip to Content
Merck
All Photos(1)

Key Documents

P7923

Sigma-Aldrich

Phosphatase, Alkaline from bovine intestinal mucosa

buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein

Synonym(s):

Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

form

buffered aqueous glycerol solution

Quality Level

specific activity

≥4,000 DEA units/mg protein

mol wt

dimer ~160 kDa

concentration

≥10.0 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase can be used to dephosphorylate casein and other proteins. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase..
Alkaline phosphatase is usually conjugated to antibodies and other proteins in immunological techniques such as ELISA, Western blotting, and immunohistochemistry. It is routinely used to dephosphorylate proteins and nucleic acids. It may be used for protein labeling when high sensitivity is required. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase. Product P7923 has been used in a novel screening strategy for nitrilase-producing strains.
High specific activity grade recommended for antibody and protein conjugation.
The enzyme has been used to develop a highly sensitive immunoassay for determining one thousandth of an attomole (1 zeptomole) of alkaline phosphatase. It has been used in an immunoassay of proinsulin. It has also been used to pretreat ISG (immature secretory granule) membranes to study the phosphorylation of proteinacous components in it.

Biochem/physiol Actions

Alkaline phosphatase, from bovine intestinal mucosa, is most stable in the pH range 7.5-9.5. The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It requires zinc, and magnesium or calcium divalent ions for activity.
The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It is routinely used to dephosphorylate proteins and nucleic acids.
The enzyme is a glycoprotein containing approximately 12% carbohydrate (6% hexoses and 6% other neutral sugars). Each molecule of alkaline phosphatase contains four zinc atoms and four disulfide bridges. Maximal activity with alkaline phosphatase is achieved in the presence of magnesium. It catalyzes the hydrolysis of phosphate monoesters such as: p-nitrophenyl phosphate, phenyl phosphate, phenolphthalein phosphate, α-glycerol phosphate, β-glycerol phosphate, 2-phosphorylglycerate, triosephosphate, glucose-6-phosphate, glucose 1-phosphate, fructose 1-phosphate, fructose 6-phosphate, adenosine 5-phosphate adenosine 3-phosphate, phosphoenolpyruvate, and β-nicotinamide adenine dinucleotide phosphate. Arsenate, cysteine, iodine, inorganic phosphate, pyrophosphate, diisopropyl phosphate, triphenylphosphate, diisopropyl fluorophosphate, and L-phenylalanine are some of the strong inhibitors of alkaline phosphatase.

Physical properties

Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.
Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.2 The enzyme requires zinc, and magnesium or calcium divalent ions for activity.

Unit Definition

One DEA unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C. (One glycine unit is equivalent to ~3 DEA units)

Physical form

Solution in 40% glycerol containing 6 mM Tris/HCl, 6 mM MgCl2 and 0.12 mM ZnCl2, ~pH 7.6

Analysis Note

Package sizes are based on DEA units
Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D B Cook et al.
Clinical chemistry, 39(6), 965-971 (1993-06-01)
Enzyme amplification has proved to be a highly sensitive quantification technique for immunoassays. We have shown that by using a fluorescent end-point, even more sensitive enzyme amplification assays can be generated than hitherto reported. We describe some general properties of
Zhijie Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(35), 21328-21335 (2020-08-21)
Theoretical and experimental observations that catalysis enhances the diffusion of enzymes have generated exciting implications about nanoscale energy flow, molecular chemotaxis, and self-powered nanomachines. However, contradictory claims on the origin, magnitude, and consequence of this phenomenon continue to arise. To
A S Dittié et al.
The EMBO journal, 16(16), 4859-4870 (1997-08-15)
The composition of secretory granules in neuroendocrine and endocrine cells is determined by two sorting events; the first in the trans-Golgi complex (TGN), the second in the immature secretory granule (ISG). Sorting from the ISG, which may be mediated by
Qing Zhu et al.
Applied and environmental microbiology, 73(19), 6053-6057 (2007-08-07)
Nitrilases have found wide use in the pharmaceutical industry for the production of fine chemicals, and it is important to have a method by which to screen libraries of isolated or engineered nitrilase variants (including bacteria and fungi). The conventional
Yana Konokhova et al.
Skeletal muscle, 6, 10-10 (2016-02-20)
Low mitochondrial content and oxidative capacity are well-established features of locomotor muscle dysfunction, a prevalent and debilitating systemic occurrence in patients with chronic obstructive pulmonary disease (COPD). Although the exact cause is not firmly established, physical inactivity and oxidative stress

Protocols

Enzymatic Assay of Alkaline Phosphatase, Diethanolamine Assay (EC 3. 1. 3. 1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service