Skip to Content
Merck

P7539

Penicillin–Streptomycin Solution Hybri-Max

with 10,000 units penicillin and 10mg/ml streptomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for hybridoma

Synonym(s):

Pen-Strep

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.76
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell wall synthesis | interferes, protein synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

General description

Penicillin-Streptomycin is used to supplement cell culture media to control bacterial contamination.

Application

Penicillin–Streptomycin Solution Hybri-Max has been used:
  • as a supplement in Roswell Park Memorial Institute (RPMI) 1640 medium to maintain human breast cancer cell lines
  • as a component of the Dulbecco′s modified Eagle′s medium/F-12 (DMEM/F-12) to culture human chondrocytes
  • as a component of the DMEM medium to culture human hepatocyte carcinoma cells

Other Notes

20ml,100ml
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library