Skip to Content
Merck
All Photos(1)

Key Documents

P7527

Sigma-Aldrich

Prostaglandin E1

γ-irradiated, powder, BioXtra, suitable for cell culture

Synonym(s):

(11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
Beilstein:
5294062
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder
Assay:
≥60% (HPLC)

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

Assay

≥60% (HPLC)

form

powder

concentration

≥60% (HPLC)

technique(s)

cell culture | mammalian: suitable

solubility

ethanol: 1 mg/mL

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in cell culture applications for the study of prostaglandin regulated cell signaling and gene regulation.
Prostaglandin E1 has been used in maintaining mice proximal tubule, and NRK-52E, a rat renal epithelial cell line.

Biochem/physiol Actions

Prostaglandins mediate vasodilation and vasoconstriction, the E series of prostaglandin is a highly effective vasodilator. It is known to efficiently cause erythema. Prostaglandin E1 is considered as a potent pyretic factor, and acts through cerebral ventricles. Prostaglandin E1 is useful to prolong cardiovascular function in new born suffering from ductus-dependant congenital heart disease until surgery.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-term prostaglandin E1 infusion for newborns with critical congenital heart disease
Aykanat A, et al.
Pediatric Cardiology, 37(1), 131-134 (2016)
Naturally produced crystals obtained from kidney stones are less injurious to renal tubular epithelial cells than synthetic crystals
Escobar C, et al.
BJU International, 100(4), 891-897 (2007)
The Prostaglandins (2013)
Stephen A Davis et al.
Nature chemical biology, 11(7), 481-487 (2015-06-02)
Drugs that act more promiscuously provide fewer routes for the emergence of resistant mutants. This benefit, however, often comes at the cost of serious off-target and dose-limiting toxicities. The classic example is the antifungal amphotericin B (AmB), which has evaded
Signe E Cremer et al.
PloS one, 14(11), e0224891-e0224891 (2019-11-14)
Domestic dogs share the same environment as humans, and they represent a valuable animal model to study naturally-occurring human disease. Platelet proteomics holds promise for the discovery of biomarkers that capture the contribution of platelets to the pathophysiology of many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service