Skip to Content
Merck

P7116

Nunc® MicroWell 96 well polypropylene plates

96 well plate, polypropylene, natural, V-bottom, 500uL/well, non sterile, 120/cs

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, Nunc 249944

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
conical bottom natural wells, natural polypropylene , polypropylene
Size:
96 wells
Sterility:
non-sterile
Binding type:
non-treated surface
Feature:
lid: no, high flange design
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

conical bottom natural wells, natural polypropylene , polypropylene

sterility

non-sterile

feature

lid: no, high flange design

packaging

case of 120 ea (internal packs of 10)

manufacturer/tradename

Nunc 249944

plate L × W

128 mm × 86 mm

size

96 wells

well maximum volume

400 μm

working volume

400 μL

binding type

non-treated surface

General description

Achieve complete sample recovery with Nunc® 96-well polypropylene MicroWell Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

Application

Suitable for: Compound storage, Liquid phase assays, Library storage, Sample transport and storage, Screening, Sample collection, Combinatorial chemistry

Features and Benefits

  • Shared wall design increases the well volume.
  • Wider wells and rounded bottom improve mixing.
  • Conical bottom well for optimal sample recovery.
  • Black and white plates available for fluorescent- and luminescent-based applications.
  • Low binding polypropylene for homogeneous assays and storage.
  • Compatible with robotics and automated systems.
  • Polypropylene resin is resistant to most chemicals, solvents and alcohols.
  • Minimal binding of molecules with hydrophilic moities.
  • Non-toxic.
  • Products come without lids.

Legal Information

MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library