Skip to Content
Merck

P6902

Pheniramine maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H20N2 · C4H4O4
CAS Number:
Molecular Weight:
356.42
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
205-051-4
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCC(c1ccccc1)c2ccccn2

InChI

1S/C16H20N2.C4H4O4/c1-18(2)13-11-15(14-8-4-3-5-9-14)16-10-6-7-12-17-16;5-3(6)1-2-4(7)8/h3-10,12,15H,11,13H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

SSOXZAQUVINQSA-BTJKTKAUSA-N

Gene Information

human ... HRH1(3269)

Biochem/physiol Actions

Pheniramine maleate is a H1 histamine receptor antagonist. It is an alkylamine derivative. Structurally, pheniramine contains a chiral carbon atom with a hydrogen atom. Other substituents include the pyridyl, alkylamine and phenyl groups. It is used as an antiallergic drug for treating the common cold, respiratory allergies, urticaria, and systematic allergic reactions.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library