Skip to Content
Merck
All Photos(1)

Key Documents

P6532

Sigma-Aldrich

Prostaglandin E2

γ-irradiated, powder, BioXtra, suitable for cell culture

Synonym(s):

(5Z,11α,13E,15S)-11,15-Dihydroxy-9-oxoprosta-5,13-dienoic acid, Dinoprostone, PGE2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Beilstein:
4709356
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
42020658
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

powder

potency

0.25-100 ng/mL

technique(s)

cell culture | mammalian: suitable

solubility

ethanol: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@@H]1CC([C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/[C@@H](O)CCCCC)=O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-17,19,21,23H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,19+/m0/s1

InChI key

XEYBRNLFEZDVAW-ARSRFYASSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in cell culture applications for the study of prostaglandin regulated cell signaling and gene regulation.
Prostaglandin E2 has been used to determine its effects on collagen gel contraction mediated by alveolar cell carcinoma cell line A549.

Biochem/physiol Actions

Most biologically active prostaglandin.
Most biologically active prostaglandin. PGE2 induces cervical ripening and parturition; mediates bradykinin-induced vasodilation; regulates adenylyl cyclase. Tumor cells that over-express cyclooxygenase 2 display increased invasiveness, angiogenesis, and resistance to apoptosis that may be due to the PGE2-induced expression of angiogenic factors and stabilization of the anti-apoptotic protein, survivin.The effect of PGE2 on the immune system is mixed. It inhibits T cell activation in vitro, suggesting it is an immunosuppressant. However, in vivo, it appears to effect expansion of the Th17 subset and differentiation of the Th1 subset of T helper cells, marking it as an immunoactivator.

Physical form

powder -20 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of type I collagenous gels by alveolar epithelial cells
Umino T, et al.
American Journal of Respiratory Cell and Molecular Biology, 22(6), 702-707 (2000)
Takayuki Nakagawa
Hearing research, 276(1-2), 27-33 (2011-02-08)
Prostaglandins are one of the major groups of chemical mediators in the mammalian body. Among prostaglandins, prostaglandin E2 (PGE2) is the most abundant prostanoid in humans and involved in regulating many different fundamental biological functions. PGE2 signaling is mediated by
Sanjiv Dhingra et al.
Circulation, 128(11 Suppl 1), S69-S78 (2013-10-18)
Allogeneic mesenchymal stem cells (MSCs) were immunoprivileged early after cardiac implantation and improved heart function in preclinical and clinical studies. However, long-term preclinical studies demonstrated that allogeneic MSCs lost their immunoprivilege and were rejected in the injured myocardium, resulting in
Katrin D Mayer-Barber et al.
Nature, 511(7507), 99-103 (2014-07-06)
Tuberculosis remains second only to HIV/AIDS as the leading cause of mortality worldwide due to a single infectious agent. Despite chemotherapy, the global tuberculosis epidemic has intensified because of HIV co-infection, the lack of an effective vaccine and the emergence
José L Maravillas-Montero et al.
Journal of immunology (Baltimore, Md. : 1950), 194(1), 29-33 (2014-11-21)
Chemokines are chemotactic cytokines that direct the traffic of leukocytes and other cells in the body. Chemokines bind to G protein-coupled receptors expressed on target cells to initiate signaling cascades and induce chemotaxis. Although the cognate receptors of most chemokines

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service