Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NB.22
UNSPSC Code:
41105300
Pricing and availability is not currently available.
General description
Precast gels contain 4.0% agarose in 1X TBE buffer with 0.5 μg/ml ethidium bromide.DNA agarose gels contain 1× TBE buffer (0.089 M Tris-borate, pH 8.3, 2 mM EDTA) and 0.5 mg/ml ethidium bromide. RNA agarose gels contain 1× MOPS (20 mM MOPS, 5 mM sodium acetate, 1 mM EDTA and 1 mM EGTA, pH 7.0) Quality
Application
Precast agarose gels are designed for high resolution separation of nucleic acids, producing sharp DNA and RNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays that help eliminate contamination from equipment and handling. Gels can be electrophoresed right in the plastic tray.
TBE - agarose gels are available in two common concentrations (1% and 4%) for DNA electrophoresis. Ethidium bromide (0.5 μg/ml) is included in the TBE-agarose gels for easy visualization. TBE-agarose gels are available in 8, 20 and 24 well formats. The separation range for the 1.0% gel is 500-1000 bp and 8-1000 bp for the 4% gel.
MOPS - agarose gels (1.25%) are available without denaturants (e.g. formaldehyde or glyoxal) for the analysis of total RNA, in vitro RNA transcripts, and Northern blotting. RNA electrophoresis usually does not interfere with staining. If significant secondary structure is suspected, the RNA should be denatured by a method listed in the product insert. The separation range for the 1.25% MOPS gel is 0.25-10 kb.
TBE - agarose gels are available in two common concentrations (1% and 4%) for DNA electrophoresis. Ethidium bromide (0.5 μg/ml) is included in the TBE-agarose gels for easy visualization. TBE-agarose gels are available in 8, 20 and 24 well formats. The separation range for the 1.0% gel is 500-1000 bp and 8-1000 bp for the 4% gel.
MOPS - agarose gels (1.25%) are available without denaturants (e.g. formaldehyde or glyoxal) for the analysis of total RNA, in vitro RNA transcripts, and Northern blotting. RNA electrophoresis usually does not interfere with staining. If significant secondary structure is suspected, the RNA should be denatured by a method listed in the product insert. The separation range for the 1.25% MOPS gel is 0.25-10 kb.
Suitable for separation of DNA fragments ranging from 8 – 1000 base pairs.
Packaging
Package of 20 individually foil-wrapped gels.
Physical form
Gel size: 6.0 × 9.5 cm, 5.5 mm thick
Cassette size: 6.8 × 10.2 cm container tray; fits common horizontal electrophoresis units
Sample format: 8-well (portrait), 20-well (landscape), 24-well (2 × 12 well portrait)
Cassette size: 6.8 × 10.2 cm container tray; fits common horizontal electrophoresis units
Sample format: 8-well (portrait), 20-well (landscape), 24-well (2 × 12 well portrait)
Analysis Note
No detected DNase or RNase activity
Other Notes
DNA agarose gels contain 1× TBE buffer (0.089 M Tris-borate, pH 8.3, 2 mM EDTA) and 0.5 mg/ml ethidium bromide.
RNA agarose gels contain 1× MOPS (20 mM MOPS, 5 mM sodium acetate, 1 mM EDTA and 1 mM EGTA, pH 7.0)
RNA agarose gels contain 1× MOPS (20 mM MOPS, 5 mM sodium acetate, 1 mM EDTA and 1 mM EGTA, pH 7.0)
Precast gels contain 4.0% agarose in 1X TBE buffer with 0.5 μg/ml ethidium bromide.
1 of 1
This Item | |||
|---|---|---|---|
| description 24 wells (2 × 12-well portrait orientation) | description 24 wells (2 × 12-well portrait orientation) | description 8-well (portrait orientation) | description 8-well (portrait orientation) |
| storage temp. room temp | storage temp. room temp | storage temp. room temp | storage temp. room temp |
| composition
| composition - | composition - | composition - |
| grade Molecular Biology | grade Molecular Biology | grade Molecular Biology | grade Molecular Biology |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
| form gel form | form gel form | form gel form | form gel form |
0.5 mg/ml ethidium bromide agarose gel | - | - | - |
Still not finding the right product?
Explore all of our products under Precast Agarose Gels
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



